Skip to Content
Merck
All Photos(1)

Key Documents

07-3230

Sigma-Aldrich

Diethyl phthalate

SAJ special grade, ≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-(CO2C2H5)2
CAS Number:
Molecular Weight:
222.24
Beilstein:
1912500
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

grade

SAJ special grade

vapor density

7.66 (vs air)

vapor pressure

1 mmHg ( 100 °C)

Assay

≥98.0%

form

liquid

autoignition temp.

854 °F

expl. lim.

0.75 %, 187 °F

availability

available only in Japan

refractive index

n20/D 1.502 (lit.)

bp

298-299 °C (lit.)

mp

−3 °C (lit.)

density

1.12 g/mL at 25 °C (lit.)

application(s)

microbiology

SMILES string

CCOC(=O)c1ccccc1C(=O)OCC

InChI

1S/C12H14O4/c1-3-15-11(13)9-7-5-6-8-10(9)12(14)16-4-2/h5-8H,3-4H2,1-2H3

InChI key

FLKPEMZONWLCSK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

338.0 °F - closed cup

Flash Point(C)

170 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kashappa-Goud H Desai et al.
Journal of controlled release : official journal of the Controlled Release Society, 165(1), 62-74 (2012-10-30)
Herein, we describe the detailed development of a simple and effective method to microencapsulate vaccine antigens in poly(lactic-co-glycolic acid) (PLGA) by simple mixing of preformed active self-microencapsulating (SM) PLGA microspheres in a low concentration aqueous antigen solution at modest temperature
Raphael J Witorsch et al.
Critical reviews in toxicology, 40 Suppl 3, 1-30 (2010-10-12)
This article reviews laboratory and epidemiological research into the endocrine disruptive effects of components of personal care products, namely, phthalate esters, parabens, ultraviolet (UV) filters, polycyclic musks, and antimicrobials. High doses of phthalates in utero can produce “phthalate syndrome,” demasculinizing
Seungmin Na et al.
Ultrasonics sonochemistry, 19(5), 1094-1098 (2012-04-03)
This study investigated the degradation of diethyl phthalate (DEP) by sonolytic, photolytic and sonophotolytic processes. Two types of UV lamps, UVC (254 nm) and VUV (185 nm+254 nm), were combined with ultrasound (283 kHz). The pseudo-first order degradation rate constants
Seungmin Na et al.
Journal of environmental management, 101, 104-110 (2012-03-13)
Ultrasound (US) combined with ultraviolet (UV) irradiation and a titanium dioxide (TiO(2)) catalyst was used to effectively remove diethyl phthalate (DEP) from aqueous solutions. Single (sonolysis, photolysis, photocatalysis) and combined (sonophotolysis, sonophotocatalysis) processes were performed to confirm the synergistic effects
A M Api
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 39(2), 97-108 (2001-03-27)
Diethyl phthalate (DEP; CAS No. 84-66-2) has many industrial uses, as a solvent and vehicle for fragrance and cosmetic ingredients and subsequent skin contact. This review focuses on its safety in use as a solvent and vehicle for fragrance and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service