Skip to Content
Merck
All Photos(1)

Key Documents

524972

Sigma-Aldrich

Diethyl phthalate

Diethyl phthalate

99.5%

Synonym(s):

1,2-Benzodioic acid diethyl ester, 1,2-Diethyl benzene-1,2-dicarboxylate, Diethyl 1,2-benzenedicarboxylate, Ethyl phthalate, Neantine, o-Benzenedicarboxylic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-(CO2C2H5)2
CAS Number:
Molecular Weight:
222.24
Beilstein:
1912500
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
bp:
298-299 °C (lit.)
vapor pressure:
1 mmHg ( 100 °C)
Pricing and availability is not currently available.

vapor density

7.66 (vs air)

vapor pressure

1 mmHg ( 100 °C)

Assay

99.5%

form

liquid

autoignition temp.

854 °F

expl. lim.

0.75 %, 187 °F

impurities

≤0.18% water

color

APHA: ≤15

refractive index

n20/D 1.502 (lit.)

bp

298-299 °C (lit.)

mp

−3 °C (lit.)

density

1.12 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCOC(=O)c1ccccc1C(=O)OCC

InChI

1S/C12H14O4/c1-3-15-11(13)9-7-5-6-8-10(9)12(14)16-4-2/h5-8H,3-4H2,1-2H3

InChI key

FLKPEMZONWLCSK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Diethyl phthalate (DEP) is a phthalic anhydride ester (PAE) that is commonly used as a solvent in cosmetics and also as a plasticizer.[1]

Application

Diethyl phthalate can be used as a solvent for the antimony reduction of aluminum chloride-phosphorus trichloride-alkyl chloride complexes to form the corresponding alkylphosphonous dichlorides.[2] It can undergo condensation with o-phenylenediamine in the presence of sodium hydride to form dibenzo (b,f)(1,4)diazocine-6,11(5H, 12H)-dione.[3]

Other Notes

Plasticizer.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

338.0 °F - closed cup

Flash Point(C)

170 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The synthesis of alkylphosphonous dichlorides from aluminum chloride?phosphorus trichloride?alkyl chloride complexes.
Perry B J, et al.
Canadian Journal of Chemistry, 41(9), 2299-2302 (1963)
Diazocine chemistry. V. Synthesis and rearrangement of dibenzo [b, f][1, 4] diazocine-6, 11 (5H, 12H)-dione.
Paudler W W and Zeiler A G
The Journal of Organic Chemistry, 34(7), 2138-2140 (1969)
Raphael J Witorsch et al.
Critical reviews in toxicology, 40 Suppl 3, 1-30 (2010-10-12)
This article reviews laboratory and epidemiological research into the endocrine disruptive effects of components of personal care products, namely, phthalate esters, parabens, ultraviolet (UV) filters, polycyclic musks, and antimicrobials. High doses of phthalates in utero can produce “phthalate syndrome,” demasculinizing
Kashappa-Goud H Desai et al.
Journal of controlled release : official journal of the Controlled Release Society, 165(1), 62-74 (2012-10-30)
Herein, we describe the detailed development of a simple and effective method to microencapsulate vaccine antigens in poly(lactic-co-glycolic acid) (PLGA) by simple mixing of preformed active self-microencapsulating (SM) PLGA microspheres in a low concentration aqueous antigen solution at modest temperature
Ying Guo et al.
Environmental science & technology, 45(8), 3788-3794 (2011-03-26)
Because of volatilization and leaching from their application in consumer and personal care products, phthalate esters are ubiquitous contaminants in the indoor environment. In this study, we measured concentrations and profiles of 9 phthalate esters in indoor dust samples collected

Reviews

Active Filters

  1. 1 Ratings-only review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service