Skip to Content
Merck
All Photos(1)

Key Documents

03-1150

Sigma-Aldrich

Benzoic acid

SAJ first grade, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COOH
CAS Number:
Molecular Weight:
122.12
Beilstein:
636131
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

SAJ first grade

vapor density

4.21 (vs air)

vapor pressure

10 mmHg ( 132 °C)

Assay

≥99.5%

form

crystalline

autoignition temp.

1061 °F

availability

available only in Japan

bp

249 °C (lit.)

mp

121-125 °C (lit.)

density

1.32 g/cm3 at 20 °C

SMILES string

OC(=O)c1ccccc1

InChI

1S/C7H6O2/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H,8,9)

InChI key

WPYMKLBDIGXBTP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper-mediated C-H/C-H biaryl coupling of benzoic acid derivatives and 1,3-azoles.
Mayuko Nishino et al.
Angewandte Chemie (International ed. in English), 52(16), 4457-4461 (2013-03-21)
Ly Dieu Tran et al.
Journal of the American Chemical Society, 134(44), 18237-18240 (2012-10-30)
An auxiliary-assisted, copper catalyzed or promoted sulfenylation of benzoic acid derivative β-C-H bonds and benzylamine derivative γ-C-H bonds has been developed. The method employs disulfide reagents, copper(II) acetate, and DMSO solvent at 90-130 °C. Application of this methodology to the
Wanchun Xiang et al.
ChemSusChem, 6(2), 256-260 (2013-01-25)
Sensing the sun: Incorporation of a cyanomethyl benzoic acid electron acceptor into donor-π-acceptor sensitizers for dye-sensitized-solar cell is shown to lead to devices with improved conversion efficiency when compared with more widely used cyanoacetic acid acceptor.
Renbo Wei et al.
Macromolecular rapid communications, 34(4), 330-334 (2013-01-03)
A new approach is developed to fabricate highly oriented mono-domain LCE nano/microstructures through micro-molding in capillaries. Gratings and microwires as two typical examples are fabricated and characterized by polarizing optical microscopy, optical microscopy, and scanning electron microscopy. The gratings with
G C Tremblay et al.
Pharmacology & therapeutics, 60(1), 63-90 (1993-10-01)
Detoxification of sodium benzoate by elimination as a conjugate with glycine, a nonessential amino acid, provides a pathway for the disposal of waste nitrogen. Since 1979, sodium benzoate has been widely used in the therapeutic regimen to combat ammonia toxicity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service