Skip to Content
Merck
All Photos(1)

Key Documents

33047

Sigma-Aldrich

Benzoic acid

puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)

Synonym(s):

Benzenecarboxylic acid, Carboxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COOH
CAS Number:
Molecular Weight:
122.12
Beilstein:
636131
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39023903
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

reag. Ph. Eur.

vapor density

4.21 (vs air)

vapor pressure

10 mmHg ( 132 °C)

Assay

≥99.9% (alkalimetric)

form

crystalline

autoignition temp.

1061 °F

impurities

readily oxidisable substances, in accordance
≤0.002% S-compounds (as S)
≤0.005% halogen compounds (as Cl)
≤0.005% insoluble in methanol

ign. residue

≤0.005% (as SO4)

bp

249 °C (lit.)

mp

121-125 °C (lit.)

solubility

water: soluble (2.9 g/l at 25 °C)

anion traces

sulfate (SO42-): ≤20 mg/kg

cation traces

Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Pb: ≤2 mg/kg
Zn: ≤5 mg/kg

functional group

carboxylic acid
phenyl

SMILES string

OC(=O)c1ccccc1

InChI

1S/C7H6O2/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H,8,9)

InChI key

WPYMKLBDIGXBTP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzoic acid is an organic aromatic monocarboxylic acid. It occurs in the form of colorless leaflets or needles. It reacts with hydrogenating reagents to afford hexahydrobenzoic acid. On decomposition (by heating) in the presence of lime or alkali, it affords benzene and carbon dioxide. It can be synthesized by the cobalt or manganese catalyzed atmospheric oxidation of toluene.

Application

Benzoic acid may be employed as a standard in the quantitative and calorimetric studies. It may be employed as an intermediate in the synthesis of following products:
  • paints
  • pigments
  • varnish
  • wetting agents
  • aroma compounds
  • benzoyl chloride
  • benzotrichloride

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eagleson M.
Concise Encyclopedia Chemistry, 122-122 (1994)
Renbo Wei et al.
Macromolecular rapid communications, 34(4), 330-334 (2013-01-03)
A new approach is developed to fabricate highly oriented mono-domain LCE nano/microstructures through micro-molding in capillaries. Gratings and microwires as two typical examples are fabricated and characterized by polarizing optical microscopy, optical microscopy, and scanning electron microscopy. The gratings with
Copper-mediated C-H/C-H biaryl coupling of benzoic acid derivatives and 1,3-azoles.
Mayuko Nishino et al.
Angewandte Chemie (International ed. in English), 52(16), 4457-4461 (2013-03-21)
Ly Dieu Tran et al.
Journal of the American Chemical Society, 134(44), 18237-18240 (2012-10-30)
An auxiliary-assisted, copper catalyzed or promoted sulfenylation of benzoic acid derivative β-C-H bonds and benzylamine derivative γ-C-H bonds has been developed. The method employs disulfide reagents, copper(II) acetate, and DMSO solvent at 90-130 °C. Application of this methodology to the
Wanchun Xiang et al.
ChemSusChem, 6(2), 256-260 (2013-01-25)
Sensing the sun: Incorporation of a cyanomethyl benzoic acid electron acceptor into donor-π-acceptor sensitizers for dye-sensitized-solar cell is shown to lead to devices with improved conversion efficiency when compared with more widely used cyanoacetic acid acceptor.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service