Skip to Content
Merck
All Photos(1)

Key Documents

01-5220

Sigma-Aldrich

Ammonium thiocyanate solution

0.1 M, 1/10 N

Synonym(s):

Ammonium rhodanide solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4SCN
CAS Number:
Molecular Weight:
76.12
Beilstein:
3595135
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

form

liquid

availability

available only in Japan

concentration

0.1 M
1/10 N

SMILES string

N.SC#N

InChI

1S/CHNS.H3N/c2-1-3;/h3H;1H3

InChI key

SOIFLUNRINLCBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaolian Gu et al.
The Journal of investigative dermatology, 135(8), 2077-2083 (2015-04-02)
Epigenetic modifications by DNA methylation are associated with a wide range of diseases. Previous studies in psoriasis have concentrated on epigenetic changes in immune cells or in total skin biopsies that include stromal-associated changes. In order to improve our understanding
Mark A Zottola
Journal of molecular graphics & modelling, 28(2), 183-186 (2009-07-25)
Cyanide (CN) is considered to be a terrorist chemical weapon due to its ready availability in multi-kilogram quantities and multi-modal means of intoxication. The body uses the sulfur transferase enzyme rhodanese to detoxify cyanide via conversion of cyanide to thiocyanate.
G Almanzar et al.
Journal of immunological methods, 387(1-2), 36-42 (2012-10-02)
Antibody avidity, defined as the strength of binding between antibody and antigen, represents a functional measure of affinity maturation of antibodies. Determination of the antibody avidity is usually performed separating high and low avidity antibodies by dissociating agents, but measurement
Javad Azizian et al.
Molecular diversity, 6(3-4), 223-226 (2004-04-08)
A new one-pot four component procedure for synthesis of densely functionalized pyrroles using commercially available ninhydrin with phosphorane intermediates produced in the reaction between triphenylphosphine, ammonium thiocyanate (or ammonium acetate) and various dialkyl acetylenedicarboxylates was developed.
Hucheng Zhang et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 71(5), 1927-1931 (2008-09-02)
The polymer-ammonium complexes are an important class of proton conducting polymer electrolytes. In this work, poly(ethylene oxide) (PEO)-NH(4)SCN electrolytes were prepared over a large range of the salt content, and their FT-IR spectra were measured at room temperature. Based on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service