Skip to Content
Merck
All Photos(3)

Key Documents

221260

Sigma-Aldrich

Ammonium iron(III) sulfate dodecahydrate

ACS reagent, 99%

Synonym(s):

Ammonium ferric sulfate dodecahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4Fe(SO4)2 · 12H2O
CAS Number:
Molecular Weight:
482.19
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24

grade

ACS reagent

Assay

98.5-102.0% (ACS specification)
99%

form

crystals

reaction suitability

reagent type: catalyst
core: iron

impurities

≤0.01% insolubles
≤0.05% not NH4OH pptd.

pH

1 (25 °C, 482.2 g/L)

mp

39-41 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%
nitrate (NO3-): ≤0.01%

cation traces

Cu: ≤0.003%
Fe2+:, passes test (limit about 0.001%)
Zn: ≤0.003%

storage temp.

2-8°C

SMILES string

N.O.O.O.O.O.O.O.O.O.O.O.O.[Fe+3].OS([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/Fe.H3N.2H2O4S.12H2O/c;;2*1-5(2,3)4;;;;;;;;;;;;/h;1H3;2*(H2,1,2,3,4);12*1H2/q+3;;;;;;;;;;;;;;;/p-3

InChI key

LCPUDZUWZDSKMX-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Ammonium ferric sulfate dodecahydrate may be used as a catalyst to synthesize:
  • Isoamyl acetate from acetic acid and isoamyl alcohol.
  • 2-Amino-5-(5-nitro-2-thienyl)-1,3,4-thiadiazole from 5-nitrothiophene-2-carboxaldehyde thiosemicarbazone via oxidative cyclization.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Antituberculosis agents VIII: Synthesis and in vitro antimycobacterial activity of alkyl a-[5-(5-nitro-2-thienyl)-1, 3, 4-thiadiazole-2-ylthio] acetates.
Foroumadi A, et al.
Il Farmaco (Societa Chimica Italiana : 1989), 58(11), 1073-1076 (2003)
Mossbauer-Effect Study of the Ferroelectric Transitions in Potassium Ferrocyanide Trihydrate and Ferric Ammonium Sulfate Dodecahydrate.
Gleason TG and Walker JC.
Physical Review, 188(2), 893-893 (1969)
Study on the Catalytic Synthesis of Isoamyl Acetate Catalyzed by Ammonlum Ferric Sulface Dodecahydrate
Ying Z, et al.
Contemporary Chemical Industry (2006)
Contemporary Chemical Industry null
Tomasz Kleszczewski et al.
Journal of pharmaceutical and biomedical analysis, 29(4), 755-759 (2002-07-03)
A method for spectrophotometric determination of L-ascorbic acid (vitamin C) in tissues by flow injection analysis is reported. The procedure is based on the oxidation of analyse with iron(III) and 2,2'-dipyridyl. Concentrations of vitamin C in the range of 0.5-20

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service