11276905001
Roche
Interferon-γ, mouse (mIFN-γ)
recombinant (E. coli)
Synonym(s):
mIFN
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
Recommended Products
biological source
mouse
Quality Level
recombinant
expressed in E. coli
Assay
>95% (SDS-PAGE)
form
solution
specific activity
>5*10^6 U/mg
packaging
pkg of 100,000 U (20 μg, 1 ml)
manufacturer/tradename
Roche
storage temp.
−20°C
Related Categories
General description
(mIFN- γ , NIH, reference standard, Gg 02–901-533, at least the same specific activity (EC 50 ) compared to the indicated standard is guaranteed.
IFN-γ (interferon-γ) is produced by natural killer T cells, macrophages, Th1 CD4 T cells and activated CD8 T cells and the production increases with age.
Recombinant, Interferon-γ, mouse (mlFN-γ) is produced in E. coli and purified by standard chromatographic techniques.
Primary structure: One polypeptide chain (131 amino acids) identical to mouse, natural IFN- ? (133 amino acids), but lacking the last two amino acids at the carboxy-terminus and not glycosylated. Glycosylation is not essential for biological activity.
Primary structure: One polypeptide chain (131 amino acids) identical to mouse, natural IFN- ? (133 amino acids), but lacking the last two amino acids at the carboxy-terminus and not glycosylated. Glycosylation is not essential for biological activity.
Specificity
Species specificity: mouse
Application
Interferon-γ, mouse (mIFN-γ) has been used in cell culture.
Recombinant mouse interferon-γ is a valuable tool for the study of IFN-γ (interferon-γ) actions in mouse model systems.
Biochem/physiol Actions
Interferon-γ (IFNγ) participates in cell-mediated immunity and has pro-inflammatory effects on immunocytes and target tissue. Its functions vary and is tissue specific. IFNγ induces T-cell-mediated immune response by increasing MHC-I expression on target cells and acts through IFNγ receptor, which is expressed on both immune and non-immune cells. IFNγ is associated with the development of colitis and Sjögren Syndrome.
Quality
- Purity: >95% (SDS-PAGE)
- Endotoxin level: <0.1 EU/μg (LAL-test), <10 EU/ml (LAL-test)
Sequence
Chain Length 131 AA
The primary structure of recombinant Interferon-γ, mouse (mIFN-γ) is identical to that of natural, mouse IFN-γ (one polypeptide chain, 131 amino acids), however, recombinant mIFN-γ lacks the last two amino acids at the C-terminus and is not glycosylated. Glycosylation is not essential for biological activity.
Unit Definition
EC 50 definition : The amount of mIFN- γ that is required to produce equivalent antiviral activity to that expressed by 1 unit of the NIH IFN-γ reference standard (Gg 02–901-533) (L cells-EMC virus/cytopathic effect) (1 unit equals =0.2 ng/ml).
EC50 definition: One unit is defined as the amount of mlFN-γ that is required to produce equivalent antiviral activity to that expressed by 1 unit of the NIH IFN-γ reference standard (Gg 02-901-533). (L cells-EMC virus/cytopathic effect) (1 unit equals ≤0.2 ng/ml).
Physical form
Solution, filtered through 0.2 μm pore size membrane, 100 000 U/ml (20 μg/ml) in PBS (phosphate buffered saline) and BSA (bovine serum albumin),1 mg/ml [purity of BSA: 98%, endotoxin (LAL): < 1 EU/mg BSA].
Preparation Note
Working solution: Dilute the concentrated mlFN-γ solution (100,000 U/ml) with PBS or culture medium containing BSA, 1 mg/ml (0.1%) or serum, 1 to 10%.
Storage conditions (working solution): -15 to -25 °C
Store the solution in aliquots at -15 to -25 °C.
Avoid repeated freezing and thawing.
Storage conditions (working solution): -15 to -25 °C
Store the solution in aliquots at -15 to -25 °C.
Avoid repeated freezing and thawing.
Other Notes
For life science research only. Not for use in diagnostic procedures.
For life science research only. Not for use in diagnostic procedures.
1 EU corresponds to 0.1 ng.
1 EU corresponds to 0.1 ng.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Interferon-? derived from cytotoxic lymphocytes directly enhances their motility and cytotoxicity.
Bhat P, et al.
Cell Death & Disease, 8(6), e2836-e2836 (2017)
The Rac-specific exchange factors Dock1 and Dock5 are dispensable for the establishment of the glomerular filtration barrier in vivo.
Laurin M, et al.
Small Gtpases, 4(4), 221-230 (2013)
Interferon-gamma deficiency protects against aging-related goblet cell loss.
Volpe E A, et al.
Oncotarget, 7(40), 64605-64605 (2016)
Natividad R Fuentes et al.
Journal of lipid research, 62, 100026-100026 (2021-01-31)
Epidermal growth factor receptor (EGFR) signaling drives the formation of many types of cancer, including colon cancer. Docosahexaenoic acid (DHA, 22∶6Δ4,7,10,13,16,19), a chemoprotective long-chain n-3 polyunsaturated fatty acid suppresses EGFR signaling. However, the mechanism underlying this phenotype remains unclear. Therefore
The mTORC2/Akt/NF?B pathway-mediated activation of TRPC6 participates in adriamycin-induced podocyte apoptosis.
Zhang H T, et al.
Cellular Physiology and Biochemistry, 40(5), 1079-1093 (2016)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service