11040596001
Roche
Interferon-γ, human (hIFN-γ)
from human, 95% (SDS-PAGE), solution, >2 x 10^7 U/mg
Synonym(s):
SYBR, qPCR
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352207
Recommended Products
biological source
human
Quality Level
recombinant
expressed in E. coli
description
recombinant (E. coli)
Assay
95% (SDS-PAGE)
form
solution
potency
<0.05 ng/mL EC50
specific activity
>2 x 10^7 U/mg
mol wt
16,700 Da
packaging
pkg of 100,000 U (5 μg, 1 ml)
manufacturer/tradename
Roche
storage condition
avoid repeated freeze/thaw cycles
impurities
<0.1 EU/μgtested (LAL test)
UniProt accession no.
storage temp.
−20°C
Gene Information
human ... IFNG(3458)
General description
IFN-γ is produced by T-lymphocytes stimulated by antigen or by T-cell mitogens. Under nondenaturating conditions its molecular weight values range from 32,000 to 73,000 indicating that recombinant human IFN-γ exists as a dimer or higher oligomers.
Specificity
Human IFN-γ is effective on human cells, but not on mouse or rat cells.
Application
Interferon-γ, human (hIFN-γ) has been used to stimulate mesenchymal stem cells (MSCs). It has also been used as a supplement Dulbecco′s modified Eagle′s medium/Ham′s medium for the cultivation of human colorectal carcinoma cell line T84 cells.
Interferon-γ, human (hIFN-γ), can be used to investigate IFN-γ activities in human cell systems.
Biochem/physiol Actions
A broad range of biological activities has been attributed to IFN-γ (e.g., the establishment of the antiviral state, immunoregulatory functions, antiproliferative effects and inhibition of cell growth). The anti-proliferative effects of IFN-γ are superior to those of either IFN-α or IFN-β. Growth inhibition is dependent on cell type, dose, and length of exposure. One of IFN-γ′s primary functions might be as an immunoregulatory agent: IFN-γ induces MHC antigens on many cells, Fc-receptors on monocytes and macrophages, IL-2 receptors on T-cells, enhances activity of macrophages, polymorphonuclear leukocytes, T-lymphocytes, and NK-cells (MAF), and is also involved in the regulation of B-cells.
Interferon-γ (IFN-γ) is an essential component of acquired immunity. A broad range of biological activities has been attributed to IFN-γ (e.g., the establishment of the antiviral state, immunoregulatory functions, antiproliferative effects and inhibition of cell growth). The anti-proliferative effects of IFN-γ are superior to those of either IFN-α or IFN-β. Growth inhibition is dependent on cell type, dose, and length of exposure. One of IFN-γ′s primary functions might be as an immunoregulatory agent: IFN-γ induces MHC antigens on many cells, Fc-receptors on monocytes and macrophages, IL-2 receptors on T-cells, enhances activity of macrophages, polymorphonuclear leukocytes, T-lymphocytes, and NK-cells (MAF), and is also involved in the regulation of B-cells.
Quality
The raw material used for this preparation was tested for HBs antigen and for the presence of antibodies to HIV-1, HIV-2, and HCV found to be negative.
Endotoxin level: <0.1EU/μg (LAL-test), <10EU/ml (LAL-test)
Note: 1EU corresponds to 0.1ng
Endotoxin level: <0.1EU/μg (LAL-test), <10EU/ml (LAL-test)
Note: 1EU corresponds to 0.1ng
Sequence
Chain Length 143 AA
The primary structure of recombinant human IFN-γ (143 amino acids) is identical to that of natural human IFN-γ (146 amino acids), however, recombinant IFN-γ has three amino acids less and is not glycosylated. Glycosylation is not essential for biological activity.
Unit Definition
EC50 definition: The amount of hIFN-γ that is required to produce equivalent antiviral activity to that expressed by 1 unit of the NIH IFN-γ reference standard (Gg 23– 901-530) (WISH cells-EMC virus/cytopathic effect) (1 unit equals ≤0.05 ng/ml).
Physical form
100,000U/ml or 1,000,000U/ml of Human IFN-γ in 0.1M phosphate buffer saline (PBS) (pH 7.0), 2.5% sucrose (w/v), and 2.5% human serum albumin (HSA) (w/v), filtered through a 0.2μm pore size membrane.
Preparation Note
Working solution: Solvent is recommended in double-distilled water.
Storage conditions (working solution): Stability in culture: Activity is present for at least 18 hours in culture medium.
Storage conditions (working solution): Stability in culture: Activity is present for at least 18 hours in culture medium.
Other Notes
EC 50 : <0.05ng/ml (hIFN-γ , NIH, reference standard, Gg 23–901-530)
For life science research only. Not for use in diagnostic procedures.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
No data available
Flash Point(C)
No data available
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Ilona E Keller et al.
Scientific reports, 5, 10230-10230 (2015-05-20)
Impaired immune function contributes to the development of chronic obstructive pulmonary disease (COPD). Disease progression is further exacerbated by pathogen infections due to impaired immune responses. Elimination of infected cells is achieved by cytotoxic CD8(+) T cells that are activated
IFNgamma modulates human immunoglobulin receptor expression in lipoaspirate-derived mesenchymal stem cells
Laksmitawati DR, et al.
Medical Journal of Indonesia, 23(3), 127-132 (2014)
Reza Norian et al.
Veterinary research forum : an international quarterly journal, 6(4), 265-271 (2016-03-15)
This in vitro study was conducted to evaluate lymphocyte blastogenic and cytokine production by bovine peripheral blood mononuclear cells (PBMCs) stimulated with phytohemagglutinin (PHA), pokeweed mitogen (PWM) and concanavalin A (Con A) mitogens, by using tetrazolium salt and ELISA tests
Permeability analyses and three-dimensional imaging of interferon gamma-induced barrier disintegration in intestinal organoids
Bardenbacher M, et al.
Stem Cell Research, 35, 101383-101383 (2019)
Anti-proliferative effect of IFN-gamma in immune regulation. II. IFN-gamma inhibits the proliferation of murine bone marrow cells stimulated with IL-3, IL-4, or granulocyte-macrophage colony-stimulating factor
Gajewski TF, et al.
Journal of immunology (Baltimore, Md. : 1950), 141, 2635-2642 (2019)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service