Skip to Content
Merck
All Photos(1)

Key Documents

TX1389

Supelco

2,2,4-Trimethylpentane

OmniSolv®

Synonym(s):

Isooctane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2C(CH3)3
CAS Number:
Molecular Weight:
114.23
Beilstein:
1696876
MDL number:
UNSPSC Code:
12191502
EC Index Number:
208-759-1

vapor density

3.9 (vs air)

Quality Level

vapor pressure

41 mmHg ( 21 °C)

product line

OmniSolv®

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

745 °F

expl. lim.

6 %

technique(s)

HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable

impurities

≤0.01% Water (H2O)
≤5 ng/L Capillary ECD responsive substances (as PCNB)

evapn. residue

≤1 ppm

color

APHA: ≤10

refractive index

n20/D 1.391 (lit.)

pH

7

bp

98-99 °C (lit.)

mp

−107 °C (lit.)

transition temp

flash point -12 °C

solubility

0.56 mg/L

density

0.692 g/mL at 25 °C (lit.)

UV cutoff

≤200 nm

UV absorption

λ: 200 nm Amax: ≤1.00
λ: 220 nm Amax: ≤0.05
λ: 230 nm Amax: ≤0.02
λ: 250 nm Amax: ≤0.005
λ: 270 nm Amax: ≤0.005
λ: 300 nm Amax: ≤0.005

shipped in

ambient

storage temp.

room temp

SMILES string

CC(C)CC(C)(C)C

InChI

1S/C8H18/c1-7(2)6-8(3,4)5/h7H,6H2,1-5H3

InChI key

NHTMVDHEPJAVLT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

OmniSolv®
TX1389P-1 is packaged in poly coated glass bottles

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Assay (GC)99.5% minCapillary ECD responsive substances (as PCNB)5ng/L maxColor (APHA)10 maxEvaporation residue1ppm maxFiltered through 0.2 µm filterTo pass testFluorescence (as quinine base)250ppt maxFormClear liquidInfrared SpectrumConforms to standardRefractive index (at 20°C)UV Abs. at 200 nm1.00AU maxUV Abs. at 220 nm0.05AU maxUV Abs. at 230 nm0.02AU maxUV Abs. at 250 nm0.005AU maxUV Abs. at 270 nm0.005AU maxUV Abs. at 300 nm0.005AU maxUV Cut-off200nm maxWater (H2O)0.01% max

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service