Skip to Content
Merck
All Photos(1)

Key Documents

SX0420

Supelco

Sodium chloride

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Halite, NaCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-598-3
NACRES:
NA.21
Assay:
≥99.0% (silver nitrate titration)
solubility:
358 g/L
Pricing and availability is not currently available.

Agency

meets USP/NF

Quality Level

200
300

vapor pressure

1.3 hPa ( 865 °C)

Assay

≥99.0% (silver nitrate titration)

impurities

≤0.005% Insoluble matter

pH

4.5-7.0 (20 °C, 100 g/L in H2O)

bp

1461 °C/1013 hPa

mp

801 °C (lit.)

solubility

358 g/L

anion traces

bromide (Br-): ≤0.01%
chlorate, nitrate (as NO3-): ≤0.003%
iodide (I-): ≤0.002%
phosphate (PO43-): ≤5 ppm
sulfate (SO42-): ≤0.004%

cation traces

Ba: ≤0.001%, passes test
Ca: ≤0.002%
Fe: ≤2 ppm
K: ≤0.005%
Mg: ≤0.001%
heavy metals: ≤5 ppm

shipped in

ambient

storage temp.

room temp

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Analysis Note

Assay (argentometric)99.0% minBarium (Ba)Passes test (about 0.001%)Bromide (Br)0.01% maxCalcium (Ca)0.002% maxChlorate and nitrate (as NO3)0.003% maxHeavy Metals (by ICP-OES)5ppm maxInsoluble matter0.005% maxIodide (I)0.002% maxIron (Fe)2ppm maxMagnesium (Mg)0.001% maxPhosphate (PO4)5ppm maxPotassium (K)0.005% maxSulfate (SO4)0.004% maxpH of a 5% solution at 25°C5.0-9.0

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service