SF006
ESGRO Complete Accutase
The ESGRO Complete Accutase is a cell detachment solution of proteolytic & collagenolytic enzymes, qualified for use for the detachment of mouse embryonic stem cells cultured in serum-free conditions with ESGRO Complete Clonal Grade Medium.
Synonym(s):
Stem Cell Tested Accutase
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352207
eCl@ss:
32160801
NACRES:
NA.75
Recommended Products
Quality Level
form
liquid
manufacturer/tradename
Chemicon®
technique(s)
cell culture | stem cell: suitable
input
sample type: mouse embryonic stem cell(s)
sample type induced pluripotent stem cell(s)
shipped in
dry ice
General description
ESGRO Complete™ Accutase is a cell detachment solution of proteolytic and collagenolytic enzymes that has been qualified for use for the detachment of mouse embryonic stem cells cultured in serum-free conditions with ESGRO Complete Clonal Grade Medium (Cat. No. SF001-500). Accutase does not contain mammalian or bacterial derived products.
Application
Cell Detachment:
1. Thaw Accutase® to room temperature.
2. Wash plate, flask or beads with sterile PBS.
3. Add Accutase to culture dish or flask using aseptic procedures at 10 mL per 75 cm2 surface area.
4. Return culture to 37°C incubator and allow cells to detach (5-10 minutes).
5. Count cells and passage as usual. No additional washes or enzyme inhibitors are required.
1. Thaw Accutase® to room temperature.
2. Wash plate, flask or beads with sterile PBS.
3. Add Accutase to culture dish or flask using aseptic procedures at 10 mL per 75 cm2 surface area.
4. Return culture to 37°C incubator and allow cells to detach (5-10 minutes).
5. Count cells and passage as usual. No additional washes or enzyme inhibitors are required.
Physical form
Frozen sterile liquid, ready to use formulation. Each lot is tested for Sterility (by USP membrane filtration method), enzymatic activity (tested with synthetic chromagenic tetrapeptides) and cell detachment from tissue culture plastic.
1X Accutase® enzymes in Dulbecco′s PBS containing 0.5 mM EDTA•4Na and 3 mg/L Phenol Red.
1X Accutase® enzymes in Dulbecco′s PBS containing 0.5 mM EDTA•4Na and 3 mg/L Phenol Red.
Storage and Stability
Stable when stored at -20°C. Refer to lot expiration date on label. Recommended storage upon receipt is -20°C. After thawing, Accutase® may be stored for up to 2 months at 4°C. DO NOT STORE AT ROOM TEMPERATURE.
Legal Information
Accutase is a registered trademark of Innovative Cell Technologies, Inc.
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
cOmplete is a trademark of Roche
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Lina Dahl et al.
PloS one, 3(4), e2025-e2025 (2008-04-24)
The molecular mechanisms regulating the expansion of the hematopoietic system including hematopoietic stem cells (HSCs) in the fetal liver during embryonic development are largely unknown. The LIM-homeobox gene Lhx2 is a candidate regulator of fetal hematopoiesis since it is expressed
Darren A Cusanovich et al.
Cell, 174(5), 1309-1324 (2018-08-07)
We applied a combinatorial indexing assay, sci-ATAC-seq, to profile genome-wide chromatin accessibility in ∼100,000 single cells from 13 adult mouse tissues. We identify 85 distinct patterns of chromatin accessibility, most of which can be assigned to cell types, and ∼400,000
Thangaselvam Muthusamy et al.
Stem cell reports, 3(1), 169-184 (2014-07-30)
We describe the use of a characteristic blue fluorescence to identify and isolate pluripotent human embryonic stem cells and human-induced pluripotent stem cells. The blue fluorescence emission (450-500 nm) is readily observed by fluorescence microscopy and correlates with the expression of
Na Suo et al.
Glia, 67(7), 1320-1332 (2019-03-01)
Oligodendrocytes (OLs) are the myelinating glia of the central nervous system. Injury to OLs causes myelin loss. In demyelinating diseases, such as multiple sclerosis, the remyelination is hindered principally due to a failure of the oligodendrocyte precursor cells (OPCs) to
Clara Jana-Lui Busch et al.
Bio-protocol, 9(14) (2020-01-08)
Alveolar macrophages (AM) are tissue-resident macrophages that colonize the lung around birth and can self-maintain long-term in an adult organism without contribution of monocytes. AM are located in the pulmonary alveoli and can be harvested by washing the lungs using
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service