Skip to Content
Merck
All Photos(1)

Key Documents

NS170

Sigma-Aldrich

Multi Species Phosphorylated Neurofilament ELISA Kit

measures and quantifies pNF-H levels in serum or plasma

Synonym(s):

200 kDa neurofilament protein, Phosphorylated Neurofilament heavy polypeptide, pNF-H

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

Product Name

Phosphorylated Neurofilament (pNF-H) Sandwich ELISA Kit, The Phosphorylated Neurofilament H (pNF-H) kit is a sandwich enzyme immunoassay (EIA), which measures pNF-H.

Quality Level

species reactivity

pig, feline, rat, human, bovine, mouse

manufacturer/tradename

Chemicon®

assay range

sensitivity: 0.0293-15 ng/mL
(Range of Detection)

sensitivity: 0.0585 ng/mL

technique(s)

ELISA: suitable

NCBI accession no.

UniProt accession no.

application(s)

research use

detection method

colorimetric

shipped in

dry ice

Application

Research Category
Neuroscience
The Phosphorylated Neurofilament H (pNF-H) kit is a sandwich enzyme immunoassay (EIA), which measures pNF-H.
Used to detect/quantify: Phosphorylated Neurofilament (pNF-H)

Packaging

96 wells

Components

Immunoplate Pre-coated with Chicken anti-pNF-H Polyclonal Antibody: (Part No. 2006263). One 96-well immunoplate pre-coated with chicken anti- pNF-H polyclonal antibody sealed in a foil pouch.

Rabbit anti- pNF-H Polyclonal Antibody: (Part No. 2006919). One vial (100 μL) of rabbit anti- pNF-H polyclonal antibody.

Goat anti-Rabbit, Alkaline Phosphatase Conjugated Antibody: (Part No. 2006917). One vial (10 μL) of goat anti-rabbit alkaline phosphatase conjugate.

Bovine pNF-H Standard: (Part No. 2006918). One vial (5.0 ng) of bovine pNF-H standard.

20X TBS: (Part No. 2004913). One bottle (50 mL) of 20X TBS.

20% Tween® 20: (Part No. 2004915). One vial (3 mL) of 20% Tween-20.

Elisa Assay Diluent: (Part No. 2006921) One bottle (30 mL) of Elisa assay diluent containg 0.05% Kathon as a preservative, provided in a ready to use format.

pNPP Buffer: (Part No. 2006261). One bottle (12 mL) of pNPP buffer for use with 50X p-NPP, provided in a ready to use format.

50X pNPP: (Part No. 2006262). One vial (250 μL) of 50X pNPP (p-nitrophenyl phosphatase) alkaline phosphatase substrate.

Plate Covers - Two Plate Covers.

Storage and Stability

Prior to use maintain the kit components at 2°-8°C and -20°C, as indicated, until expiration date.

1. Wash Buffer - After dilution maintain the Wash Solution at 2°-8°C for up to one month or until expiration date.

2. Tween® 20 - Store at room temperature upon receipt of kit.

3. Elisa Assay Buffer - Aliquot as required to avoid repeated freeze-thaw cycles. After thawing, maintain the Elisa Assay Buffer at 2°-8°C for up to one month, or until expiration date.

4. Immunoplate Pre-coated with Chicken anti- pNF-H Polyclonal Antibody - Unused strip wells may be returned to the foil pouch and stored at 2°-8°C for up to one month, or until expiration date.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
TWEEN is a registered trademark of Croda International PLC

Disclaimer

For research use only. Not for use in diagnostic procedures.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Oral

Target Organs

Kidney,Liver,Blood

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Petzold et al.
Neuro-degenerative diseases, 4(2-3), 185-194 (2007-06-29)
Loss of cortical neurons is a key pathological feature in neurodegenerative dementias. Cerebrospinal fluid (CSF) neurofilaments (Nf) are a biomarker for neuronal death and axonal loss. To perform a meta-analysis to investigate the value of CSF Nf levels for the
Gerry Shaw et al.
Biochemical and biophysical research communications, 336(4), 1268-1277 (2005-09-24)
Several lines of reasoning suggest that the phosphorylated axonal form of the neurofilament subunit NF-H is likely to be released from damaged and diseased neurons in significant amounts. Detection of this protein in serum or CSF might therefore provide information
Kevin J Anderson et al.
Journal of neurotrauma, 25(9), 1079-1085 (2008-08-30)
The detection of neuron-specific proteins in blood might allow quantification of the degree of neuropathology in experimental and clinical contexts. We have been studying a novel blood biomarker of axonal injury, the heavily phosphorylated axonal form of the high molecular
P N Hoffman et al.
The Journal of cell biology, 66(2), 351-366 (1975-08-01)
This study of the slow component of axonal transport was aimed at two problems: the specific identification of polypeptides transported into the axon from the cell body, and the identification of structural polypeptides of the axoplasm. The axonal transport paradigm
A Petzold et al.
Journal of neurology, neurosurgery, and psychiatry, 75(8), 1178-1180 (2004-07-20)
To investigate whether plasma biomarkers for axonal injury and inflammation are related to loss and recovery of visual function in acute optic neuritis (ON). Eighteen patients with ON and 14 controls were investigated in a longitudinal, prospective study. Plasma phosphorylated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service