Skip to Content
Merck
All Photos(5)

Key Documents

MABN1817

Sigma-Aldrich

Anti-α-Synuclein (SNCA) Antibody

mouse monoclonal, 2F12

Synonym(s):

Alpha-synuclein, NACP, Non-A beta component of AD amyloid, Non-A4 component of amyloid precursor, Synuclein alpha-140

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

Product Name

Anti-α-Synuclein Antibody, clone 2F12, clone 2F12, from mouse

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2F12, monoclonal

species reactivity

rat, human, mouse

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... SNCA(6622)

General description

Alpha-synuclein (UniProt P37840; also known as NACP, Non-A beta component of AD amyloid, Non-A4 component of amyloid precursor, Synuclein alpha-140) is encoded by the SNCA (also known as NACP, PARK1, PARK4) gene (Gene ID 6622) in human. Pathological aggregates are common features of many neurodegenerative diseases, such as tau neurofibrillary tangles (NFTs) in Alzheimer’s disease (AD) and frontotemporal degeneration, and α-synuclein (α-syn or αS) Lewy bodies (LBs) in Parkinson’s disease (PD) and dementia with LBs (DLB). Alpha-synuclein is a phospholipid-binding protein concentrated in presynaptic terminals where it promotes SNARE complex formation and modulates synaptic functions. Alpha-synuclein is the major component of pathologic inclusions that characterize PD, DLB, and multiple system atrophy (MSA). Research shows that αS exists not only as unfolded monomers, but in large part also as multimers, principally as ~60 kDa tetramers composed of four N-acetylated αS, that assume α-helical conformation and resist aggregation. PD-causing αS missense mutations are found to shift cellular αS from tetramers/multimers to monomers, indicating that decreased α-helical tetramers and increased unfolded monomers initiate pathogenesis. In addition, both casein kinase-1 (CK-1) and CK-2 can catalyze the phosphorylation of αS on Ser129, and Ser129-phosphorylated αS is found in αS inclusions.

Specificity

Clone 2F12 reacted with both monomeric and aggregated forms of alpha-synuclein of human, mouse, and rat species. Clone 2F12 detected both wild-type alpha-synuclein and fPD mutants (Dettmer, U., et al. (2015). Nat. Commun. 6:7314; Dettmer, U., et al. (2013). J. Biol. Chem. 288(9):6371-6385).

Immunogen

Purified human erythrocyte α-synuclein.

Application

Anti-α-Synuclein, clone 2F12, Cat. No. MABN1817, is a highly specific mouse monoclonal antibody that targets α-synuclein and has been tested in ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected α-synuclein in human prostate cancer, cerebral cortex, and kidney tissue sections.

ELISA Analysis: A representative lot (0.4 µL in 30 µL buffer/well for coating) captured recombinant human α-synuclein (0.2-40 ng/mL) in a sandwich ELISA application utilizing clone SOY1 (Cat. No. MABN1818; preconjugated with Sulfo tag) as the detection antibody (Courtesy of Tim Bartels, Ph.D., Brigham and Women′s Hospital, Boston, MA, U.S.A.).

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot immunostained primary mouse cortical neurons (Courtesy of Tim Bartels, Ph.D., Brigham and Women′s Hospital, Boston, MA, U.S.A.).

Immunohistochemistry Analysis: A 1:11,110 dilution from a representative lot immunostained Lewy bodies (LBs) in striatum tissue sections from Parkinson′s diseased (PD) human brain (Courtesy of Tim Bartels, Ph.D., Brigham and Women′s Hospital, Boston, MA, U.S.A.).

Immunoprecipitation Analysis: 4 µL from a representative lot immunoprecipitated α-synuclein from 50 µg of HEL human erythroleukemia cell lysate (Courtesy of Tim Bartels, Ph.D., Brigham and Women′s Hospital, Boston, MA, U.S.A.).

ELISA Analysis: A representative lot captured both endogenous α-synuclein (αS) from human cortical homogenate, as well the exogenously expressed wild type and familial PD (fPD) αS mutants (A30P, E46K, H50Q, G51D, A53T) from sytosolic extracts of transfected M17D human neuroblastoma cells in a sandwich ELISA application utilizing clone SOY1 (Cat. No. MABN1818; preconjugated with Sulfo tag) as the detection antibody (Dettmer, U., et al. (2015). Nat. Commun. 6:7314).

ELISA Analysis: A representative lot captured both pre-aggregated fibrillar recombinant α-synuclein as well as partially purified Lewy bodies (LBs) from a DLB (dementia with LBs) patient with or without prior sample denaturing by boiling with 2% SDS in a sandwich ELISA application utilizing clone SOY1 (Cat. No. MABN1818; preconjugated with Sulfo tag) as the detection antibody (Dettmer, U., et al. (2015). Nat. Commun. 6:7314).

Immunocytochemistry Analysis: A representative lot detected cytosolic localization of endogenous rat α-synuclein (αS) and exogenously overexpressed human αS by fluorescent immunocytochemistry staining of 4% paraformaldehyde-fixed, 0.25% Triton X-100-permeabilized primary rat neurons and transfected M17D human neuroblastoma cells (Dettmer, U., et al. (2015). Nat. Commun. 6:7314).

Western Blotting Analysis: A representative lot detected monomeric α-synuclein (αS) as well as αS multimers (αS60, αS80 and αS100) in extract from disuccinimidyl glutarate (DSG) cross-linked mouse brain bits, human iPSCs (both S A53T mutant and corrected isogenic line) and ESCs (both wild-type and genetically engineered isogenic αS E46K line). A significantly reduced αS60 level was seen with A53T and E46K mutants (Dettmer, U., et al. (2015). Nat. Commun. 6:7314).

Western Blotting Analysis: A representative lot detected monomeric α-synuclein (αS) as well as αS multimers (αS60, αS80 and αS100) in cytosolic extracts from disuccinimidyl glutarate (DSG) cross-linked primary rat neurons, as well as human HEL erythroid leukemia and M17D neuroblastoma cells (Dettmer, U., et al. (2013). J. Biol. Chem. 288(9):6371-6385).
Research Category
Neuroscience

Quality

Evalulated by Western Blotting in human fetal brain tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected α-synuclein in 10 µg of human fetal brain tissue lysate.

Target description

~14.5 kDa observed. 14.46 kDa (human isoform 1; NACP140), 14.49/14.52 kDa (mouse/rat isoform 1) calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cristina Román-Vendrell et al.
Frontiers in neuroscience, 15, 639414-639414 (2021-02-23)
α-Synuclein is a presynaptic protein that regulates synaptic vesicle trafficking under physiological conditions. However, in several neurodegenerative diseases, including Parkinson's disease, dementia with Lewy bodies, and multiple system atrophy, α-synuclein accumulates throughout the neuron, including at synapses, leading to altered
Anwesha Sanyal et al.
Journal of molecular biology, 431(12), 2266-2282 (2019-04-30)
During disease, cells experience various stresses that manifest as an accumulation of misfolded proteins and eventually lead to cell death. To combat this stress, cells activate a pathway called unfolded protein response that functions to maintain endoplasmic reticulum (ER) homeostasis
Evan Casalino et al.
SLAS discovery : advancing life sciences R & D, 27(6), 349-357 (2022-05-18)
Small-molecule high-throughput screening (HTS) campaigns have frequently been used to identify lead molecules that can alter expression of disease-relevant proteins in cell-based assays. However, most cell-based HTS assays require short compound exposure periods to avoid toxicity and ensure that compounds
Thibaut Imberdis et al.
Methods in molecular biology (Clifton, N.J.), 1948, 77-91 (2019-02-17)
β-Sheet-rich aggregates of α-synuclein (αS) are the hallmark neuropathology of Parkinson's disease (PD) and related synucleinopathies, whereas the native conformations of αS in healthy cells are under debate. Cross-linking analyses in intact cells detect a large portion of endogenous αS
Ulf Dettmer et al.
Human molecular genetics, 26(18), 3466-3481 (2017-09-16)
α-Synuclein (αS) forms round cytoplasmic inclusions in Parkinson's disease (PD) and dementia with Lewy bodies (DLB). Evidence suggests a physiological function of αS in vesicle trafficking and release. In contrast to earlier tenets, recent work indicates that αS normally exists

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service