Skip to Content
Merck
All Photos(1)

Key Documents

MABE1799

Sigma-Aldrich

Anti-Nrf2 Antibody, clone 103

clone 103, from rat

Synonym(s):

Nuclear factor erythroid 2-related factor 2, NF-E2-related factor 2, NFE2-related factor 2, Nuclear factor, erythroid derived 2, like 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

103, monoclonal

species reactivity

rat, mouse

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... Nfe2L2(18024)

General description

Nuclear factor erythroid 2-related factor 2 (UniProt: Q60795; also known as Nrf2, NF-E2-related factor 2, NFE2-related factor 2, Nuclear factor, erythroid derived 2, like 2) is encoded by the Nfe2l2 (also known as Nrf-2, Nrf2) gene (Gene ID: 18024) in murine species. Nrf2 is a cap n collar basic-region leucine zipper transcription factor that is involved in adaptation of cells to oxidative stress and electrophiles. It bind to antioxidant response (ARE) elements in the promoter regions of target genes and induces a set of cytoprotective genes that are involved in the synthesis of reduced glutathione (GSH), repair of oxidized protein thiols, scavenging of reactive oxygen species, and drug metabolism. It is cytosolic under unstressed conditions, but translocates into the nucleus upon induction by electrophilic agents. Under normal unstressed conditions it is rapidly turned over in a Keap-1 dependent manner through Cul3-Rbx1 ubiquitination and proteasomal degradation. Phosphorylation of Ser40 by protein kinase C in response to oxidative stress dissociates Nrf2 from its Keap-1 and promotes its translocation into the nucleus. It is widely expressed with highest expression in liver, skeletal muscle, luminal cells of the stomach and intestine, lining of the bronchi and alveoli, and in renal tubules; followed by heart, spleen, testis and brain. Nrf2 is constitutively upregulated in many tumors due to somatic mutations in the Keap1 or Nrf2 genes. Tumors overexpressing Nrf2 exhibit increased resistance to chemotherapeutic drugs and higher rates of proliferation. (Ref.: Hayes, JD., and Ashford, MLJ (2012). Cell Metab. 16(2): 139-141).

Specificity

Clone 103 detects Nrf2 in Keap-1 knockout cells. It targets an epitope within 179 amino acids in the internal region.

Immunogen

GST-tagged recombinant fragment corresponding to 179 amino acids from internal region of murine Nuclear factor erythroid 2-related factor 2.

Application

Anti-Nrf2, clone 103 , Cat. No. MABE1799, is a highly specific rat monoclonal antibody that targets Nuclear factor erythroid 2-related factor 2 and has been tested in Immunohistochemistry and Western Blotting.
Immunohistochemistry Analysis: A representative lot detected Nrf2 in Immunohistochemistry applications (Saito, R., et. al. (2015). Mol Cell Biol. 36(2):271-84).

Western Blotting Analysis: A representative lot detected Nrf2 in Western Blotting applications (Maruyama, A., et. al. (2008). Arch Biochem Biophys. 477(1):139-45; Kobayashi, E.H., et. al. (2016). Nat Commun. 7:11624; Taquchi, K., et. al. (2016). Toxicol Sci. 152(1):40-52; Saito, R., et. al. (2015). Mol Cell Biol. 36(2):271-84; Taquchi, K., et. al. (2014). Mol Cell Biol. 34(5):900-13).
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in Keap1-KO MEF cell lysates versus Nrf2-KO MEF cell lysates.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Nrf2 in Keap1-KO MEF cell lysates versus Nrf2-KO MEF cell lysates.

Target description

~67 kDa observed; 66.90 kDa calculated Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carmen Rodríguez-García et al.
Molecular nutrition & food research, 66(18), e2200082-e2200082 (2022-07-19)
To compare the effects of three high-fat diets (HFDs) based on coconut, sunflower, or extra virgin olive oils (EVOOs) on adipose tissue, metabolism, and inflammation. Mice are fed for 16 weeks on their respective HFD. HFD based on coconut oil produces
Yuening Liu et al.
The Journal of biological chemistry, 297(1), 100850-100850 (2021-06-05)
Reperfusion therapy, the standard treatment for acute myocardial infarction, can trigger necrotic death of cardiomyocytes and provoke ischemia/reperfusion (I/R) injury. However, signaling pathways that regulate cardiomyocyte necrosis remain largely unknown. Our recent genome-wide RNAi screen has identified a potential necrosis
Ye Tian et al.
PloS one, 18(1), e0279964-e0279964 (2023-01-07)
Sepsis associated encephalopathy (SAE) is a common but poorly understood complication during sepsis. Currently, there are no preventive or therapeutic agents available for this neurological disorder. The present study was designed to determine the potential protective effects of β-patchoulene (β-PAE)
Sirinat Pengnet et al.
Frontiers in pharmacology, 13, 1049818-1049818 (2023-01-03)
Background: Excessive fructose consumption causes hepatic lipid accumulation via increased triglyceride (TG) synthesis, leading to the development and progression of non-alcoholic fatty liver disease (NALFD). Naringin, a flavanone glycoside found in citrus fruit, has antioxidant and hypolipidemic properties. Therefore, the
Greg A Timblin et al.
Nature metabolism, 3(5), 618-635 (2021-05-26)
Macrophages generate mitochondrial reactive oxygen species and mitochondrial reactive electrophilic species as antimicrobials during Toll-like receptor (TLR)-dependent inflammatory responses. Whether mitochondrial stress caused by these molecules impacts macrophage function is unknown. Here, we demonstrate that both pharmacologically driven and lipopolysaccharide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service