Skip to Content
Merck
All Photos(2)

Key Documents

MAB3026

Sigma-Aldrich

Anti-NFκB Antibody, p65 subunit, active subunit, clone 12H11

clone 12H11, Chemicon®, from mouse

Synonym(s):

Rel A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

12H11, monoclonal

species reactivity

human, rabbit, rat

species reactivity (predicted by homology)

mouse

packaging

antibody small pack of 25 μg

manufacturer/tradename

Chemicon®

technique(s)

electrophoretic mobility shift assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG3

NCBI accession no.

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... NFKB1(4790)

General description

The transcription factor NFkappaB (Nuclear Factor kappa B) is involved in the expression and regulation of a number of important cellular and physiological processes such as growth, development, apoptosis, immune and inflammatory response, and activation of various viral promoters including human immunodeficiency virus long terminal repeats. NFkappaB represents a group of structurally related and evolutionarily conserved proteins related to the proto-oncogene c-Rel with five members in mammals that include Rel (cRel), RelA (p65), RelB, NFkappaB1 (p50 and its precursor p105), and NFkappaB2 (p52 and its precursor p100). NFkappaB/Rel proteins exist as homo- or heterodimers to form transcriptionally competent or repressive complexes. Although most NFkappaB dimers are activators of transcription, the p50/50 and p52/52 homodimers can repress the transcription of their target genes. The p50/p65 heterodimer of NFkappaB is the most abundant in cells.

Specificity

Recognizes an epitope overlapping the nuclear location signal (NLS) of the p65 subunit of the NFkB heterodimer. Thus it selectively binds to the activated form of NFkB (Zabel et al., 1993). The antibody can also be used for electrophoretic mobility supershift assays (EMSA). The nuclear factor kB (NFkB) is a sequence-specific DNA-binding protein (Sen & Baltimore, 1986). It is a pleiotropic transcription factor which is involved in the expression of a variety of cellular and viral genes (Lenardo & Baltimore, 1989). NFkB consists of two subunits which are named according to their molecular weight, p50 and p65 (Kawakami et al., 1988). The subunits are stabilized by a so-called inhibitory chain IkB (Baeuerle & Baltimore, 1988). In quiescent cells, NFkB resides in the cytosol in an inactive form which can be activated in vivo by treatment of cells with cytokines or protein kinase activators. In vitro, it is possible to generate the active form of NFkB by treatment with sodium deoxycholate, formadine or electrophoretic size fractionation. The active form of NFkB is a heterotetrameric protein, consisting of the two p50 and two p65 subunits (Lenardo et al., 1987). After activation NFkB translocates from the cytosol to the nucleus of the cell, binds to specific DNA sequences and initiates transcription.

Immunogen

Epitope: p65 subunit, active subunit
Peptide corresponding to human p65 coupled to BSA.

Application

Immunofluorescence:
A 1-10 μg/mL concentration of a previous lot was used in immunofluorescence.

Immunohistochemistry (paraffin sections):
A 5-10 μg/mL (APAAP) concentration of a previous lot was used in immunohistochemistry.

Immunohistochemistry (frozen sections):
A 5-10 μg/mL (APAAP) concentration of a previous lot was used in immunohistochemistry.

Immunohistochemistry:
The clone 12H11 works best in fresh frozen or acetone fixed human tissues, however some groups have had reactivity in traditional formalin fixed tissue when the tissue is of human origin. It is recommended that ABC or enhanced detection systems be employed for the best visualization in either acetone or formalin fixed tissues. The antibody reacts with human tissues best. Rat tissue will also react but at a lower affinity, and the antibody does not react with mouse, other species have not been examined. Microwave citric acid buffer or trypsin digestion antigen recovery have both been successful with 12H11 on formalin fixed tissues.

Western blot: 5-10 µg/mL (ECL)

Electrophoretic Mobility Supershift Assay:
A 0.5-1 μg/mL concentration of a previous lot was used in Supershift assay.

Flow cytometry:
A previous lot of this antibody was used in flow cytometry. Fixed cells only, acetone fixed cells.

Optimal working dilutions must be determined by end user.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
Use Anti-NFκB Antibody, p65 subunit, active subunit, clone 12H11 (Mouse Monoclonal Antibody) validated in EMSA, FC, ICC, IF, IHC, IHC(P), WB to detect NFκB also known as Rel A.

Quality

Routinely evaluated by Western Blot on PC12 lysates.

Western blot:
1:500 dilution of this lot detected NF KAPPA B, P65 on 10 μg of PC12 lysates.

Target description

65 kDa

Physical form

Format: Purified
Protein A purified
Purified mouse monoclonal IgG3 liquid in buffer containing 0.02 M Phosphate buffer, 0.25 M NaCl, pH 7.6 with 0.1% sodium azide.

Storage and Stability

Stable for 6 months at 2-8ºC from date of receipt.
Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
TNF α-treated HeLa cells, PMA and calcium ionophore-treated Jurkat cells.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Stephen A Watt et al.
The American journal of pathology, 185(9), 2354-2363 (2015-07-28)
NF-κB signaling plays a crucial role in regulating proliferation and differentiation in the epidermis. Alterations in the NF-κB pathway can lead to skin pathologies with a significant burden to human health such as psoriasis and cutaneous squamous cell carcinoma (cSCC).
Jayeeta Ghose et al.
RNA biology, 12(4), 457-477 (2015-03-12)
MicroRNA (miRNA) genes generally share many features common to those of protein coding genes. Various transcription factors (TFs) and co-regulators are also known to regulate miRNA genes. Here we identify novel p53 and NFκB p65/RelA responsive miRNAs and demonstrate that
Zhen-Peng Song et al.
Acta pharmacologica Sinica, 37(6), 753-762 (2016-05-10)
To investigate the mechanisms underlying the anti-nociceptive effect of minocycline on bone cancer pain (BCP) in rats. A rat model of BCP was established by inoculating Walker 256 mammary carcinoma cells into tibial medullary canal. Two weeks later, the rats
Novel signaling mechanisms of intracellular angiotensin II-induced NHE3 expression and activation in mouse proximal tubule cells.
Li, XC; Hopfer, U; Zhuo, JL
American Journal of Physiology: Renal Physiology null
Angela L Bush et al.
Brain research, 1203, 18-25 (2008-03-04)
The avian brainstem serves as a useful model to answer the question of how afferent activity influences the viability of target neurons. Approximately 20-30% of neurons in the avian cochlear nucleus, nucleus magnocellularis (NM) die following deafferentation (i.e., deafness produced

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service