Skip to Content
Merck
All Photos(1)

Documents

MAB3262F

Sigma-Aldrich

Anti-BrdU Antibody, clone PRB-1, FITC conjugated

clone PRB-1, Chemicon®, from mouse

Synonym(s):

BrdU

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

PRB-1, monoclonal

species reactivity (predicted by homology)

all

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable

isotype

IgG1

shipped in

wet ice

target post-translational modification

unmodified

Related Categories

Specificity

Recognizes BrdU in ssDNA, free BrdU or BrdU coupled to a protein carrier. MAB3262 also reacts with iodouridine. Does not react with thymidine.

Immunogen

Iodouridine conjugated to ovalbumin.

Application

Anti-BrdU Antibody, clone PRB-1, FITC conjugated is a Mouse Monoclonal Antibody for detection of Bromodeoxyuridine also known as BrdU & has been validated in FC.
Flow cytometry: 1:20

Optimal working dilutions must be determined by the end user.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair

Physical form

Purified immunoglobulin conjugated to fluorescein. Liquid in PBS with 0.05% sodium azide pH 7.4

Storage and Stability

Maintain at 2-8°C in undiluted aliquots up to 6 months. Protect from prolonged exposure to light.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CCN1 induces hepatic ductular reaction through integrin ?v??-mediated activation of NF-?B.
Kim, KH; Chen, CC; Alpini, G; Lau, LF
The Journal of Clinical Investigation null
Yu Yan et al.
Oncology reports, 51(2) (2023-12-15)
The radioresistance of glioma is an important cause of treatment failure and tumor aggressiveness. In the present study, under performed with linear accelerator, the effects of 0.3 and 3.0 Gy low‑dose radiation (LDR) on the proliferation and migration of C6 glioma
Yan Fan et al.
Neurotoxicity research, 38(3), 793-807 (2020-07-04)
The present study investigated the effects of forced overexpression of Phox2a/2b, two transcription factors, in the locus coeruleus (LC) of aged rats on noradrenergic and dopaminergic phenotypes in brains. Results showed that a significant increase in Phox2a/2b mRNA levels in
Maxim L Bychkov et al.
Biomedicines, 10(3) (2022-03-26)
Metastatic melanoma is a highly malignant tumor. Melanoma cells release extracellular vesicles (EVs), which contribute to the growth, metastasis, and malignancy of neighboring cells by transfer of tumor-promoting miRNAs, mRNA, and proteins. Melanoma microenvironment acidification promotes tumor progression and determines
Maria Plesa et al.
Frontiers in immunology, 12, 715848-715848 (2021-09-14)
Toll-like receptor 3 (TLR3) activation by viral infections plays a key role in promoting inflammatory immune responses that contribute to pulmonary fibrosis in chronic inflammatory respiratory diseases. Vitamin D3 has been shown to be beneficial to patients with asthma and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service