Skip to Content
Merck
All Photos(1)

Documents

IM29L

Sigma-Aldrich

Anti-PAI-1 (Ab-1) Mouse mAb (2)

lyophilized, clone 2, Calbiochem®

Synonym(s):

Anti-Plasminogen Activator Inhibitor-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

2, monoclonal

form

lyophilized

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG1

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... SERPINE1(5054)

Related Categories

General description

Purified mouse monoclonal antibody (see application references). Recognizes both active and latent forms of the ~50 kDa PAI-1 protein.
Recognizes both active and latent forms of the ~50 kDa PAI-1 protein in conditioned media from dexamethasone-treated HT-1080 cells. Sold under license of U.S. Patent 5,422,245.
This Anti-PAI-1 (Ab-1) Mouse mAb (2) is validated for use in Immunoblotting, Immunoprecipitation, Immunocytochemistry for the detection of PAI-1 (Ab-1).

Immunogen

Human
partially purified PAI-1 from conditioned medium of dexamethasone-treated HT1080 human cells

Application

Immunoblotting (1 µg/ml)

Immunoprecipitation (see application references)

Immunocytochemistry (see application references

Neutralization Studies (see application references)

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from a volatile buffer, 100 µg BSA.

Reconstitution

We recommend resuspending the lyophilized antibody with sterile phosphate buffered saline (PBS), pH 7.4, or sterile 20 mM Tris-saline (20 mM Tris containing 0.15 M NaCl), pH 7.4, to yield a final concentration of 100 µg/ml. Lyophilized antibodies should be resuspended at 4°C with occasional gentle mixing for at least 2 h.

Analysis Note

Positive Control
Conditioned media from dexamethasone-treated (1 µM) HT-1080 cells

Other Notes

Antibody should be titrated for optimal results in individual systems.
Janicke, F., et al. 1994. Cancer Research.54, 2527.
Lawrence, D. A., et al. 1994. J. Biol. Chem.269, 15223.
Nagayama, M., et al. 1994. Cancer73, 1398.
Petersen, H., et al. 1994. Cancer Res.54, 120.
Keijer, J., et al. 1991. Blood78, 401.
Blasi, F. and Verde, P., 1990. in Seminars in Cancer Biol.Vol. 1, 153.
Declerck, P. J. and Collen, D., 1990. Thrombosis Res.Supplement X 3.
Nielsen, L. S., et al. 1986. Thrombosis and Haemostasis.55, 206.

Legal Information

Sold under license of U.S. Patent 5,422,245.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service