Skip to Content
Merck
All Photos(2)

Key Documents

HNPMAG-35K

Millipore

MILLIPLEX® Human Neuropeptide Magnetic Bead Panel - Neuroscience Multiplex Assay

The analytes available for this multiplex kit are: α-MSH, β-Endorphin, Neurotensin, Oxytocin, Substance P.

Synonym(s):

Human Neuroscience Protein Multiplex Assay, Luminex® Human Neuroscience Panel, Millipore Human Neuroscience Immunoassay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

species reactivity

human

manufacturer/tradename

Milliplex®

assay range

accuracy: 98-118%
(Spike Recovery)

sensitivity: 11-479 pg/mL
(MinDC+2SD)

standard curve range: 27-20,000 pg/mL
(α-MSH)

standard curve range: 41-30,000 pg/mL
(Neurotensin)

standard curve range: 55-40,000 pg/mL
(Oxytocin)

standard curve range: 69-50,000 pg/mL
(β-Endorphine)

standard curve range: 7-5,000 pg/mL
(Substance P)

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

The central nervous system is a complex environment consisting of billions of neurons and glial support cells. These neurons use chemical signals to communicate information. Neuropeptides are neuronal signaling molecules secreted by the central and peripheral nervous systems, influencing a broad spectrum of biological functions including the regulation of metabolism, reproduction, immunity, and behavior. Understanding the role of neuropeptides in the pathogenesis of devastating neuronal diseases and behavioral disorders is fundamental. While it is understood that accurate measurement of neuropeptides is important, conventional methods, including RIAs and ELISAs, are not able to simultaneously measure multiple neuropeptides with small sample volume. MILLIPLEX® Human Neuropeptide Bead Panel is a 5-plex kit to be used for the simultaneous quantification of any or all of the following analytes in serum, plasma, cerebrospinal fluid (CSF), tissue/cell lysate, and culture supernatant samples: α-Melanocyte Stimulating Hormone (α-MSH), β-Endorphin, Neurotensin, Oxytocin, and Substance P.

The MILLIPLEX® Human Neuropeptide Bead Panel is to be used for the simultaneous quantification of the following 5 analytes in any combination: α-MSH, β-Endorphin, Neurotensin, Oxytocin, and Substance P. This kit may be used for the analysis of all or any combination of the above analytes in tissue/cell lysate, culture supernatant samples, and cerebrospinal fluid (CSF). Serum or plasma samples may also be used and will need to be extracted. An important note about this conversion panel is that the samples may be extracted using either the acetonitrile precipitation technique or the Waters 96-well HLB extraction plate. Please refer to the protocol for more information on both methods. The necessary components to perform the extractions are not included with the kit.

Panel Type: Neuroscience

Specificity

There was no or negligible cross-reactivity between the antibodies for an analyte and any of the other analytes within a panel.

Application

  • Analytes: α-MSH, β-Endorphin, Neurotensin, Oxytocin, Substance P
  • Note that this is a competitive assay.
  • Recommended Sample type: serum, plasma, tissue culture supernatant, or CSF
  • Recommended Sample dilution: extracted serum or plasma or neat CSF or culture supernatant
  • Assay Run Time: Overnight
  • Research Category: Neuroscience

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Packaging

Everything you need in a single kit.

Components

Human Neuropeptide Standard, 1 vial, lyophilized

Human Neuropeptide Quality Control 1, 1 vial, lyophilized

Human Neuropeptide Quality Control 2, 1 vial, lyophilized

Assay Buffer, 2 bottles, 30 mL ea.

Wash Buffer, 10X (0.05% Proclin), 2 bottles, 30 mL ea

Human Neuropeptide Primary Antibodies, 1 bottle, 3.5 mL

Human Neuropeptide Detection Antibodies, 1 bottle, 5.5 mL

Human Neuropeptide Matrix Solution, 1 bottle, 1.5 mL

Human Neuropeptide Antibody-Immobilized Magnetic Beads, 20X, 200 uL ea

Streptavidin-Phycoerythrin, 1 bottle, 5.5 mL

Mixing Bottle, 1 bottle

Sample Plate, 96 well format, 1 plate with 2 Foil Plate Sealers

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Other Notes

Sensitivity: Refer to kit protocol for sensitivities of individual biomarkers.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Immediate effects of spinal manipulation on nitric oxide, substance P and pain perception.
Molina-Ortega, F; Lomas-Vega, R; Hita-Contreras, F; Plaza Manzano, G; Achalandabaso et al.
Manual therapy null
Emmanuel Bäckryd et al.
Pain medicine (Malden, Mass.), 15(1), 111-119 (2013-10-15)
Pain medicine still lacks mechanism-specific biomarkers to guide diagnosis and treatment, and defective top-down modulation is an important factor in the pathophysiology of chronic pain conditions. Using modern analytical tools and advanced multivariate statistical analysis, the aim of this study
Olga V Roschina et al.
Journal of clinical medicine, 10(23) (2021-12-11)
The neuropeptides β-endorphin and oxytocin are released into the bloodstream as hormones from the pituitary gland but also have an important function as neuroregulators in the forebrain. The blood levels of both polypeptides have been shown to reflect depressive symptoms.
Ann M Manzardo et al.
American journal of medical genetics. Part A, 170(9), 2328-2333 (2016-08-16)
Prader-Willi syndrome (PWS) is a rare genetic neurodevelopmental disorder associated with maladaptive social behavior, hyperphagia and morbid obesity. Orexin A is a hypothalamic neuropeptide important as a homeostatic regulator of feeding behavior and in energy metabolism through actions in the
Estefania Simoes et al.
Frontiers in nutrition, 7, 613301-613301 (2021-03-02)
Objective: Childhood obesity is a growing concern as the World Health Organization (WHO) states that ~10% of adolescents worldwide are overweight or obese. This condition is the reflex of energy imbalance between the calories consumed and those expended. Sex-related responses

Related Content

Learn how multiplex detection of Alzheimer’s disease biomarkers, using the MILLIPLEX® Human Amyloid Beta and Tau Magnetic Bead Panel, allows for a different perspective on neurodegenerative disease research.

Learn how multiplex detection of Alzheimer’s disease biomarkers, using the MILLIPLEX® Human Amyloid Beta and Tau Magnetic Bead Panel, allows for a different perspective on neurodegenerative disease research.

Learn how multiplex detection of Alzheimer’s disease biomarkers, using the MILLIPLEX® Human Amyloid Beta and Tau Magnetic Bead Panel, allows for a different perspective on neurodegenerative disease research.

Learn how multiplex detection of Alzheimer’s disease biomarkers, using the MILLIPLEX® Human Amyloid Beta and Tau Magnetic Bead Panel, allows for a different perspective on neurodegenerative disease research.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service