Skip to Content
Merck
All Photos(1)

Key Documents

HKI5MAG-99K

Millipore

MILLIPLEX® Human Kidney Injury Magnetic Bead Panel 5 - Toxicity Multiplex Assay

The analytes available for this multiplex kit are: α-1-Microglobulin, Collagen IV, Lipocalin-2/NGAL, Osteoactivin, TIMP-1, Uromodulin.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

species reactivity

human

manufacturer/tradename

Milliplex®

assay range

accuracy: 62-90%
intra-assay cv: <10
sensitivity: 0.05-0.56 ng/mL
(MinDC+2SD)

standard curve range: 0.05-50 ng/mL
(Lipocalin-2/NGAL)

standard curve range: 0.05-50 ng/mL
(Uromodulin)

standard curve range: 0.10-100 ng/mL
(TIMP-1)

standard curve range: 0.20-200 ng/mL
(Osteoactivin)

standard curve range: 0.24-250 ng/mL
(α-1-Microglobulin)

standard curve range: 0.49-500 ng/mL
(Collagen IV)

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

Maintenance of physiological balance in the body is attributed in part to normal kidney function. Altered kidney function due to injury, drug toxicity or kidney failure may be life threatening. Toxicity is the leading cause of drug failure, so research is expanding in search of more sensitive, rapid methods to determine organ-specific damage as quickly as possible. Traditionally, blood urea nitrogen (BUN) and serum creatinine tests have been used to determine drug-induced damage to the kidney. These tests only detect kidney damage after it begins and do not allow for the elucidation of where in the kidney the damage has occurred. Specificity and earlier detection of kidney injury is vital, and this area of research is expanding in search of more sensitive, rapid methods for determining the specific area of damage in kidney.

The MILLIPLEX® portfolio provides valuable research assays to investigate multiple biomarkers of kidney injury in human serum and plasma samples using the Luminex® xMAP® instrument platform.

The MILLIPLEX® Human Kidney Injury Bead Panel 5 contains all the components necessary to measure the following six biomarkers in any combination using Luminex® xMAP® technology: α-1-Microglobulin, Collagen IV, Lipocalin-2/NGAL, Osteoactivin, TIMP-1, Uromodulin. The kit uses a 96-well format, contains a lyophilized standard cocktail, two quality controls and can measure up to 38 serum or plasma samples in duplicate.

Panel Type: Toxicity

Specificity

There was no or negligible cross-reactivity between the antibodies for an analyte and any of the other analytes within a panel.

Application

  • Analytes: α-1-Microglobulin, Collagen IV, Lipocalin-2/NGAL, Osteoactivin, TIMP-1, Uromodulin
  • Recommended Sample type: serum and plasma
  • Recommended Sample dilution: 1:75 in kit Assay Buffer. Customers should determine the optimal dilution factor for their samples. Our recommendations are based on serum and plasma samples from normal subjects
  • Assay Run Time: Overnight
  • Research Category: Inflammation & Immunology

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Packaging

Everything you need in a single kit.

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Other Notes

Please contact Technical Service for linearity of dilution.
Sensitivity: Please see kit protocol for individual assay sensitivities.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Multiplex kidney toxicity assays detect nephrotoxicity biomarkers in small samples, minimizing time and costs for toxicity screening.

Multiplex kidney toxicity assays detect nephrotoxicity biomarkers in small samples, minimizing time and costs for toxicity screening.

Multiplex kidney toxicity assays detect nephrotoxicity biomarkers in small samples, minimizing time and costs for toxicity screening.

Multiplex kidney toxicity assays detect nephrotoxicity biomarkers in small samples, minimizing time and costs for toxicity screening.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service