Skip to Content
Merck
All Photos(2)

Key Documents

RECYTMAG-65K

Millipore

MILLIPLEX® Rat Cytokine/Chemokine Magnetic Bead Panel - Immunology Multiplex Assay

Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in rat serum, plasma and cell culture samples.

Synonym(s):

luminex rat cytokine chemokine growth factor multiplex assay, luminex rat pro inflammatory anti inflammatory cytokine panel, millipore rat inflammation multiplex kit, rat interleukin immunoassay panel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

species reactivity

rat

manufacturer/tradename

Milliplex®

assay range

accuracy: 80-131%
standard curve range: 0.2-1,000 pg/mL
(EGF)

standard curve range: 12.2-50,000 pg/mL
(GM-CSF, IL-1α, IL-2, IL-12 (p70), IL-18)

standard curve range: 14.6-60,000 pg/mL
(Leptin, IFNγ, GRO/KC/CINC-1)

standard curve range: 2.4-10,000 pg/mL
(MIP-1α, IL-1β, IP-10, Fractalkine, TNFα)

standard curve range: 24.4-100,000 pg/mL
(LIX, MIP-2)

standard curve range: 29.3-120,000 pg/mL
(MCP-1)

standard curve range: 4.9-20,000 pg/mL
(G-CSF, Eotaxin, IL-4, IL-13, IL-5, VEGF, RANTES)

standard curve range: 7.3-30,000 pg/mL
(IL-10, IL-17A)

standard curve range: 73.2-300,000 pg/mL
(IL-6)

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

The MILLIPLEX® Rat Cytokine/Chemokine Bead Panel (RECYTMAG-65K) enables you to focus on the therapeutic potential of cytokines, as well as the modulation of cytokine expression.

Panel Type: Cytokines/Chemokines

Specificity

Cross Reactivty
There is no or minimum cross-reactivity between the antibodies and any other analytes in the panel.

Application

  • Analytes: EGF, Eotaxin/CCL11, Fractalkine, G-CSF, GM-CSF, GRO/KC, IFN-γ, IL-1α, IL-1β, IL-2, IL-4, IL-5, IL-6, IL-10, IL-12 (p70), IL-13, IL-17A, IL-18, IP-10, Leptin, LIX, MCP-1, MIP-1α, MIP-2, RANTES, TNF-α, VEGF
  • Recommended Sample type: rat serum or plasma
  • Recommended Sample dilution: 1:2
  • Assay Run Time: One day

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Linkage

Replaces: RCYTOMAG-80K

Legal Information

MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ginsenoside Rd maintains adult neural stem cell proliferation during lead-impaired neurogenesis.
Wang, B; Feng, G; Tang, C; Wang, L; Cheng, H; Zhang, Y; Ma, J; Shi, M; Zhao, G
Neurological Sciences null
A guinea pig model of Zika virus infection.
Kumar M, Krause KK, Azouz F, Nakano E, Nerurkar VR.
Virology Journal null
Mariano Guardia Clausi et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 37(3), 787-800 (2016-03-18)
Neuroinflammation subsequent to developmental brain injury contributes to a wave of secondary neurodegeneration and to reactive astrogliosis that can inhibit oligodendrocyte progenitor differentiation and subsequent myelination. Here we evaluated the therapeutic efficacy of a small molecule antagonist for a TGFß
?1-Antitrypsin inhibits ischemia reperfusion-induced lung injury by reducing inflammatory response and cell death.
Gao, W; Zhao, J; Kim, H; Xu, S; Chen, M; Bai, X; Toba, H; Cho, HR; Zhang, H; Keshavjeel, S; Liu, M
The Journal of Heart and Lung Transplantation : the Official Publication of the International Society For Heart Transplantation null
Jodie L Morris et al.
PloS one, 14(12), e0226574-e0226574 (2019-12-27)
Phage therapy offers a potential alternate strategy for the treatment of peri-prosthetic joint infection (PJI), particularly where limited effective antibiotics are available. We undertook preclinical trials to investigate the therapeutic efficacy of a phage cocktail, alone and in combination with

Related Content

See how multiplex assays, such as the Luminex® bead-based multiplex immunoassay MILLIPLEX® Rat Liver Injury Panel, enable researchers to simultaneously measure multiple hepatotoxic biomarkers and cytokines to expand knowledge on drug-induced liver injury processes.

See how multiplex assays, such as the Luminex® bead-based multiplex immunoassay MILLIPLEX® Rat Liver Injury Panel, enable researchers to simultaneously measure multiple hepatotoxic biomarkers and cytokines to expand knowledge on drug-induced liver injury processes.

See how multiplex assays, such as the Luminex® bead-based multiplex immunoassay MILLIPLEX® Rat Liver Injury Panel, enable researchers to simultaneously measure multiple hepatotoxic biomarkers and cytokines to expand knowledge on drug-induced liver injury processes.

See how multiplex assays, such as the Luminex® bead-based multiplex immunoassay MILLIPLEX® Rat Liver Injury Panel, enable researchers to simultaneously measure multiple hepatotoxic biomarkers and cytokines to expand knowledge on drug-induced liver injury processes.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service