Skip to Content
Merck
All Photos(1)

Key Documents

GR11L

Sigma-Aldrich

Anti-IGF-IR (Ab-1) Mouse mAb (αIR3)

lyophilized, clone αIR3, Calbiochem®

Synonym(s):

Anti-IGF-IR, Anti-Insulin-Like Growth Factor Receptor, Anti-Insulin-Like Growth Factor Receptor, Anti-IGF-IR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
clone:
αIR3, monoclonal
application:
species reactivity:
human
citations:
11

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

αIR3, monoclonal

form

lyophilized

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG1

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... IGF1R(3480)

General description

This Anti-IGF-IR (Ab-1) Mouse mAb (αIR3) is validated for use in IF, IP, Neutralization Studies, Paraffin Sections, Immunoblotting for the detection of IGF-IR (Ab-1).

Application

Immunofluorescence (1-5 µg/ml)

Immunoprecipitation (1 µg/sample)

Neutralization Studies (1 µg/ml; see application references)

Paraffin Sections (not recommended)

Immunoblotting (not recommended)

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 20 mM ammonium bicarbonate solution, 100 µg BSA .

Reconstitution

Reconstitute with sterile phosphate buffered saline (PBS), pH 7.4, or sterile 20 mM Tris-saline (20 mM Tris containing 0.15 M NaCl), pH 7.4, to a final concentration of 100 µg/ml. Lyophilized antibodies should be resuspended at 4°C with occasional gentle mixing for at least two h. Following reconstitution, aliquot and freeze (-20°C) or refrigerate (4°C) with 0.1% sodium azide. Freezing of aliquots is recommended for long term storage of reconstituted product. Avoid freeze/thaw cycles of solutions.

Other Notes

Blocks IGF-I binding to the IGF-I receptor. May also weakly cross-react with the insulin receptor (see application references). It also inhibits the growth of MCF-7 cells in culture suggesting the IGF-I receptor may be involved in autocrine regulation of cell growth (see application references). Antibody should be titrated for optimal results in individual systems.
Roth, R. 1988. Science239, 1269.
Rohlik, Q.T., et al. 1987. Biochem. Biophy Res. Comm.149, 276.
Rosen, O.M. 1987. Science257, 1452.
Rechler, M.M., and Nissley, S.P. 1986. Hormone Res.24, 152.
Ullrich, A., et al. 1986. EMBO J.5, 2503.
Zapf, S., and Froesch, E.R. 1986. Hormone Res.24, 121.
Humbel, R.E. 1984. I. Chemistry in Li Hormonal proteins and peptides. Vol 12, Chap. 4 (Academic Press, New York, 1984).
Kull, F.C., et al.1983. J. Biol. Chem.258, 6561.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katrin Friedbichler et al.
Molecular cancer therapeutics, 13(2), 399-409 (2013-12-04)
Insulin-like growth factor (IGF) signaling is thought to play a role in the development and progression of multiple cancer types. To date, therapeutic strategies aimed at disrupting IGF signaling have largely focused on antibodies that target the IGF-I receptor (IGF-IR).
C Meier et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 19(10), 3847-3859 (1999-05-11)
Although Schwann cell precursors from early embryonic nerves die in the absence of axonal signals, Schwann cells in older nerves can survive in the absence of axons in the distal stump of transected nerves. This is crucially important, because successful
Daniela Kiepe et al.
Endocrinology, 149(10), 4901-4911 (2008-06-17)
The IGF/IGF binding protein (IGFBP) system is an important component in the hormonal regulation of longitudinal growth. Evidence from in vitro studies indicates that IGFBPs may have IGF-independent effects. We analyzed the biological activity of intact IGFBP-2 and defined carboxy-terminal
Nicole Bäumer et al.
Nature protocols, 11(1), 22-36 (2015-12-04)
Knockdown of genes by RNA interference (RNAi) in vitro requires methods of transfection or transduction, both of which have limited impact in vivo. As a virus-free approach, we chemically coupled cell surface receptors internalizing antibodies to the short interfering RNA
Roxana Schillaci et al.
British journal of haematology, 130(1), 58-66 (2005-06-29)
Chronic lymphocytic leukaemia (CLL) is characterized by the accumulation of long-lived B lymphocytes blocked in G(0/1) by impaired apoptosis. As insulin-like growth factor-I (IGF-I) is known for its antiapoptotic effects in different cell types, we investigated whether IGF-I and its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service