Skip to Content
Merck
All Photos(1)

Key Documents

DX0837

Supelco

Dichloromethane

OmniSolv® HR-GC

Synonym(s):

Methylene Chloride, Methylene chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH2Cl2
CAS Number:
Molecular Weight:
84.93
Beilstein:
1730800
MDL number:
UNSPSC Code:
12352101
EC Index Number:
200-838-9

vapor density

2.9 (vs air)

Quality Level

vapor pressure

24.45 psi ( 55 °C)
6.83 psi ( 20 °C)

product line

OmniSolv®

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

1223 °F

expl. lim.

22 %

technique(s)

gas chromatography (GC): suitable

impurities

≤0.01% Water (H2O)
≤0.2 μeq/g Titrable acid

evapn. residue

≤1 ppm

color

APHA: ≤5

refractive index

n20/D 1.424 (lit.)

pH

7 (20 °C)

bp

39.8-40 °C (lit.)

mp

-95 °C
−95 °C (lit.)

density

1.325 g/mL at 25 °C (lit.)

shipped in

ambient

storage temp.

room temp

SMILES string

ClCCl

InChI

1S/CH2Cl2/c2-1-3/h1H2

InChI key

YMWUJEATGCHHMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

OmniSolv® HR-GC
Stabilized with an alkene.

Analysis Note

AppearanceClear liquid, free from particulatesAssay (GC)99.9% minCap. ECD responsive substances as PCNB (peak)2ng/L maxCap. ECD responsive substances as PCNB (total)20ng/L maxCap. FID responsive substances as decane (peak)1µg/L maxCap. FID responsive substances as decane (total)10µg/L maxColor (APHA)5 maxEvaporation residue1ppm maxFree halogensPasses testTitrable acid0.2µeq/g maxWater (H2O)0.01% max
Evaporation Number: 1.9

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service