Skip to Content
Merck
All Photos(1)

Key Documents

CX1059

Supelco

Chloroform

OmniSolv®

Synonym(s):

Trichloromethane, Methylidyne trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CHCl3
CAS Number:
Molecular Weight:
119.38
Beilstein:
1731042
MDL number:
UNSPSC Code:
51272501
EC Index Number:
200-663-8
Assay:
≥99.8% chloroform + ethanol basis
technique(s):
HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable
bp:
60.5-61.5 °C (lit.)
vapor pressure:
160 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

4.1 (vs air)

Quality Level

vapor pressure

160 mmHg ( 20 °C)

product line

OmniSolv®

Assay

≥99.8% chloroform + ethanol basis

form

liquid

contains

0.5-1 % (v/v) ethanol as stabilizer

technique(s)

HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable

impurities

≤0.01% Water (H2O)
≤0.2 μeq/g Titrable acid
≤5 ng/L Capillary ECD responsive substances (as PCNB)

evapn. residue

≤1 ppm

color

APHA: ≤10

refractive index

n20/D 1.445 (lit.)

bp

60.5-61.5 °C (lit.)

mp

−63 °C (lit.)

solubility

8.7 g/L

density

1.492 g/mL at 25 °C (lit.)

UV cutoff

≤245 nm

UV absorption

λ: 245 nm Amax: ≤1.00
λ: 255 nm Amax: ≤0.15
λ: 260 nm Amax: ≤0.05
λ: 270 nm Amax: ≤0.02
λ: 290 nm Amax: ≤0.01

shipped in

ambient

storage temp.

room temp

SMILES string

ClC(Cl)Cl

InChI

1S/CHCl3/c2-1(3)4/h1H

InChI key

HEDRZPFGACZZDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

OmniSolv®
Stabilized with Ethanol.

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Assay (CHCl3 + C2H5OH)99.8% minCapillary ECD responsive substances (as PCNB)5ng/L maxColor (APHA)10 maxEthanol0.5-1.0%(v/v)Evaporation residue1ppm maxFiltered through 0.2 µm filterTo pass testFluorescence (as quinine base)250ppt maxFormClear liquidInfrared SpectrumConforms to standardTitrable acid0.2µeq/g maxUV Abs. at 245 nm1.00AU maxUV Abs. at 255 nm0.15AU maxUV Abs. at 260 nm0.05AU maxUV Abs. at 270 nm0.02AU maxUV Abs. at 290 nm0.01AU maxUV Cut-off245nm maxWater (H2O)0.01% max

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service