Skip to Content
Merck
All Photos(2)

Key Documents

1.02444

Supelco

Chloroform

for liquid chromatography LiChrosolv®

Synonym(s):

Trichloromethane, TCM, Methane trichloride, Methyl trichloride, Methylidyne trichloride, Trichloromethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CHCl3
CAS Number:
Molecular Weight:
119.38
Beilstein:
1731042
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-663-8
NACRES:
NA.03
Assay:
≥99.8% (GC)
technique(s):
HPLC: suitable
bp:
60.5-61.5 °C (lit.)
vapor pressure:
160 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

4.1 (vs air)

Quality Level

vapor pressure

160 mmHg ( 20 °C)

product line

LiChrosolv®

grade

isocratic

Assay

≥99.8% (GC)

form

liquid

potency

695 mg/kg LD50, oral (Rat)
>3980 mg/kg LD50, skin (Rabbit)

technique(s)

HPLC: suitable

impurities

≤0.0002 meq/g Acidity
≤0.0002 meq/g Alkalinity
≤0.01% Water

evapn. residue

≤5.0 mg/L

transmittance

255 nm, ≥70%
260 nm, ≥85%
300 nm, ≥98%

refractive index

n20/D 1.445 (lit.)

bp

60.5-61.5 °C (lit.)

mp

−63 °C (lit.)

solubility

8 g/L

density

1.492 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

ClC(Cl)Cl

InChI

1S/CHCl3/c2-1(3)4/h1H

InChI key

HEDRZPFGACZZDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Preparation and application of stimuli-responsive PET TeMs: Utilizes RAFT graft block copolymerisation of styrene and acrylic acid for the separation of water-oil emulsions, demonstrating the adaptation of chloroform in enhancing polymer properties for environmental applications (Muslimova et al., 2024).

  • Effect of Stool Sampling on a Routine Clinical Method: Describes the use of chloroform in gas chromatography-mass spectrometry for the quantification of short-chain fatty acids in stool samples, highlighting its importance in clinical diagnostics (Mahdi et al., 2024).

  • Improved lipid analysis using a 2D-LC-MS system: Discusses a novel injection procedure involving chloroform for enhanced lipid profiling, showcasing its critical role in advanced biochemical analysis (Pánczél et al., 2024).

  • Ecological Risk Assessment of Three Pesticide Additives in Soil: Addresses the application of chloroform in the remediation of contaminated soils, emphasizing its utility in environmental toxicology and soil science (Li et al., 2024).

Preparation Note

Product filtered through a 0.2 μm filter

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

LICHROSOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service