Skip to Content
Merck
All Photos(1)

Key Documents

CP01

Sigma-Aldrich

Anti-Actin (Ab-1) Mouse mAb (JLA20)

liquid, clone JLA20, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
clone:
JLA20, monoclonal
application:
citations:
218

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

JLA20, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative (actin antibody only, CP01-100UL)

species reactivity (predicted by homology)

all

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgM

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACTB(60)

General description

Anti-Actin (Ab-1), mouse monoclonal, clone JLA20, recognizes the ~ 42 kDa α-, β-, and γ isoforms of actin. It is validated for Western blotting and immunofluorescence.
Mouse monoclonal antibody generated by immunizing BALB/c mice with the specified immunogen and fusing splenocytes with NS-1 mouse myeloma cells supplied as ascites. Recognizes the ~42 kDa α-, β- and γ-isoforms of actin. Supplied with goat anti-mouse IgM conjugated to horseradish peroxidase.
Recognizes the ~42 kDa α-, β-, and γ isoforms of actin. Useful as a positive control antibody.
  • Antibody Target Gene Symbol: ACTA1, ACTB1, ACTG1
  • Target Synonym: AA959943, ACTA, Acta-2, Acts, Actsk-1, ALPHA SKELETAL ACTIN, ALPHA1 ACTIN, ASMA, CFTD, CFTD1, CFTDM, Fetal isoform of skeletal alpha-actin, G-actin, MPFD, NEM1, NEM2, NEM3, SKA, SKELETAL ALPHA-ACTIN, SKELETAL MUSCLE ACTIN
  • Entrez Gene Name: actin, alpha 1, skeletal muscle
  • Hu Entrez ID: 58, 71 (Related Antibodies: CP64)
  • Mu Entrez ID: 11459 ;-; 11465
  • Rat Entrez ID: 29437 ;-; 287876
  • Immunogen

    Chicken
    chicken cytoskeletal proteins

    Application

    Immunoblotting (1:5000-1:20000)

    Immunofluorescence (1:10000, see application references)



    Recomended dilution for secondary antibody (0.05 µg/ml, 1:2000 dilution)

    Warning

    Toxicity: Multiple Toxicity Values, refer to MSDS (O)

    Physical form

    100 µl ascites diluted in 5 mM sodium phosphate buffer, pH 7.5 and 5 µg anti-mouse IgM, HRP conjugate supplied at 100 µg/ml in 0.05 M sodium phosphate, 50% glycerol.

    Reconstitution

    Following initial thaw, aliquot ascites and freeze (-20°C).

    Analysis Note

    Positive Control
    All animal cell lines

    Other Notes

    Actin is found in all animal cells. Actin monomers have a molecular weight of ~42 kDa. For best results, use the anti-mouse IgM HRP conjugate supplied with the anti-actin antibody. Useful for normalizing protein loading. Antibodies should be titrated for optimal results in individual systems.
    Lin, J.J. et al. 1984 in Monoclonal antibodies and functional cell lines. Plenum Publishing Corp.
    Lin, J.J. 1982. Cold Spring Harbor Symp. on Quant. Biol.46, 769.
    Lin, J.J. 1981. Proc. Natl. Acad. Sci.78, 2335.
    Goldman, R.D., et al. 1979. Ann. Rev. Physiol.41, 701.
    Goldman, R., et al. (eds.) 1976. Cold Spring Harbor Conference on Cell Proliferation, Books A-C.

    Legal Information

    CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

    Not finding the right product?  

    Try our Product Selector Tool.

    Storage Class Code

    11 - Combustible Solids

    WGK

    WGK 1

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Natasa J Stojkov et al.
    Molecular human reproduction, 20(1), 77-88 (2013-07-31)
    The molecular mechanism of stress-associated reproductive dysfunction is complex and largely unknown. This study was designed to systematically analyze molecular effects of systemic in vivo blockade of α1-adrenergic receptors (α1-ADRs) on stress-induced disturbance of cAMP/cGMP signaling in testosterone-producing Leydig cells
    David L Evers et al.
    Antiviral research, 68(3), 124-134 (2005-09-29)
    We report antiviral activity against human cytomegalovirus for certain dietary flavonoids and their likely biochemical mechanisms of action. Nine out of ten evaluated flavonoids blocked HCMV replication at concentrations that were significantly lower than those producing cytotoxicity against growing or
    M C Hollander et al.
    Cell cycle (Georgetown, Tex.), 4(5), 704-709 (2005-04-23)
    Gadd45a is a p53-regulated gene whose protein product, like p53, is involved in maintenance of genome stability. Specifically, deletion of Gadd45a leads to extensive aneuploidy as a consequence of centrosome amplification and subsequent abnormal segregation of chromosomes during mitosis. S-phase
    Valentina Caracciolo et al.
    American journal of physiology. Endocrinology and metabolism, 315(4), E676-E693 (2018-03-07)
    Obesity is associated with adipose tissue inflammation that contributes to insulin resistance. Zinc finger protein 36 (Zfp36) is an mRNA-binding protein that reduces inflammation by binding to cytokine transcripts and promoting their degradation. We hypothesized that myeloid-specific deficiency of Zfp36
    Peter Mullen et al.
    Clinical cancer research : an official journal of the American Association for Cancer Research, 10(6), 2100-2108 (2004-03-26)
    We sought to identify determinants of growth response to the Raf-1-targeted antisense oligonucleotide (ASO; ISIS 5132) using a large panel of ovarian cancer cell lines. First-(ISIS 5132) and second-generation (ISIS 13650) anti-Raf 1 ASOs were compared with control oligonucleotides. Growth

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service