Skip to Content
Merck
All Photos(1)

Key Documents

CBL1337

Sigma-Aldrich

Anti-PECAM-1 Antibody, clone 390

clone 390, Chemicon®, from rat

Synonym(s):

CD31

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

390, monoclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PECAM1(5175)

General description

CD31, also known as platelet endothelial cell adhesion molecule 1 (PECAM1) is a type I integral membrane glycoprotein, and a member of the immunoglobulin superfamily of cell surface receptors. PECAM-1 is constitutively expressed on the surface of endothelial cells where it is concentrated at cell junctions. PECAM-1 is also expressed at low levels on all leukocytes and platelets. Homotypic interaction of CD31 is important in cell-cell interaction, cell-matrix interaction and signal transduction.

Application

Anti-PECAM-1 Antibody, clone 390 is an antibody against PECAM-1 for use in FC, IH & IP.
Flow Cytometry: 1 μg/10e6 cells.

Immunohistochemistry: acteone fixed frozen tissue sections

Immunoprecipitation

Blocking of cell adhesion: CBL1337 inhibits aggregation of L cells transfected with a variant form of CD31 (DeLisser, 1993)

Optimal working dilutions must be determined by the end user.

Physical form

Format: Purified
Protein A Purified mouse immunoglobulin in 100mM borate buffered saline, pH 8.0, containing no preservative.

Analysis Note

Control
Jurkat and Raji cells, tonsil tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Targeting Vascular Amyloid in Arterioles of Alzheimer Disease Transgenic Mice With Amyloid ? Protein Antibody-Coated Nanoparticles.
Poduslo JF, Hultman KL, Curran GL, Preboske GM, Chamberlain R, Marjanska M, Garwood M et al.
Journal of Neuropathology and Experimental Neurology null
Simone P Niclou et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 22(9), 3120-3128 (2008-05-23)
A NOD/Scid mouse expressing enhanced green fluorescent protein (eGFP) is described, in which human and mouse tumors marked with red fluorescent protein can be established in vivo, both at subcutaneous and orthotopic locations. Using light microscopy as well as multiphoton
Katsuhiro Hayashi et al.
The Journal of surgical research, 140(2), 165-170 (2007-04-10)
Angiogenesis is a critical step in tumor growth, progression, and metastasis. Soft tissue and bone sarcoma are resistant to most therapeutic approaches. Angiogenesis of these tumors may be an effective target. We hypothesized that we could inhibit tumor growth by
Minshu Wang et al.
ESC heart failure, 8(2), 1427-1437 (2021-01-30)
Endothelial activation and inflammatory cell infiltration have important roles in the development of cardiac fibrosis induced by renin-angiotensin system activation. NADPH oxidases (Nox proteins) are expressed in endothelial cells (ECs) and alter their function. Previous studies indicated that Nox2 in
Varun Venkataramani et al.
Cell, 185(16), 2899-2917 (2022-08-02)
Glioblastomas are incurable tumors infiltrating the brain. A subpopulation of glioblastoma cells forms a functional and therapy-resistant tumor cell network interconnected by tumor microtubes (TMs). Other subpopulations appear unconnected, and their biological role remains unclear. Here, we demonstrate that whole-brain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service