Skip to Content
Merck
All Photos(1)

Key Documents

F8402

Sigma-Aldrich

Monoclonal Anti-CD31 (PECAM-1)−FITC antibody produced in mouse

clone WM-59, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Monoclonal Anti-CD31, Anti-PECAM-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

WM-59, monoclonal

form

buffered aqueous solution

mol wt

antigen 130-140 kDa

species reactivity

human

storage condition

protect from light

technique(s)

flow cytometry: 10 μL using 1 × 106 cells

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... PECAM1(5175)

Looking for similar products? Visit Product Comparison Guide

General description

CD31 is a single chain integral membrane glycoprotein member of the immunoglobulin superfamily of cell adhesion molecules with molecular mass of 130-140kDa. It has six Ig-like loops in the extracellular domain, a trans-membrane domain including a relatively long cytoplasmic domain. It is expressed abundantly on platelets, myeloid cells, B lymphocytes, certain T-lymphocyte cell subsets, bone marrow precursor cells, NK cells and localized to the intracellular junctions in monolayers of cultured endothelial cells.
Monoclonal Anti-CD31 (PECAM-1) is derived from the WM-59 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse.

Specificity

Recognizes the CD31 (PECAM-1, endoCAM, gpIIa, hec7), human cell surface integral membrane glycoprotein. The antibody increases the rate of homotypic aggregation induced in U937 cells by TGF-β1. Its reactivity with platelets is enhanced following their washing.
5th Workshop: code No. P025

Immunogen

human cell line RC-2A, originally derived from myeloid leukemia cells.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Flow cytometry/Cell sorting (1 paper)
Monoclonal Anti-CD31 (PECAM-1)-FITC antibody has been used in
  • DAB based-immunohistochemistry
  • direct immunofluorescent staining
  • fluorescence-activated cell sorting (FACS)
  • flow cytometry

Monoclonal Anti-CD31 (PECAM-1)-FITC antibody is suitable for immunostaining of HEPC-CB2 and HEPC-CB.2 cell Lines (hematopoietic cells) and flow cytometry to evaluate the percentage of contaminating endothelia cells in characterization and decidualization of endometrial stromal cells.

Biochem/physiol Actions

CD31 is also known as PECAM-1 (Platelet endothelial cell adhesion molecule-1) binds to glycosaminoglycans (GAG). Endothelial CD31 is phosphorylated on tyrosine and serine residues. CD31 is involved in some of the interactive events taking place during cardiovascular development inflammation, thrombosis, wound healing and angiogenesis.
CD31 performs in homophilic and heterophilic cell-cell adhesion and cell signalling activities. It plays a major role in the transmigration of monocytes, neutrophils and NK cells between the endothelial cell junctions into the subendothelial matrix.

Target description

Human CD31 (PECAM-1) is expressed on platelets, endothelial cells, myeloid cells, B lymphocytes, and certain T lymphocyte subsets. CD31 (PECAM-1) functions in homophilic and heterophilic cell-cell adhesion and cell signaling activities.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Preparation Note

Prepared by conjugation to fluorescein isothiocyanate isomer I (FITC). This green dye is efficiently excited at 495 nm and emits at 525 nm.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P J Newman
Annals of the New York Academy of Sciences, 714, 165-174 (1994-04-18)
Platelet/Endothelial Cell Adhesion Molecule-1 (PECAM-1) is a 130-kDa integral membrane glycoprotein found on the surface of human platelets and leukocytes, and at the intercellular junctions of endothelial cells. PECAM-1 is a member of the immunoglobulin (Ig) gene superfamily, with six
Platelet/endothelial cell adhesion molecule-1 (CD31)-mediated cellular aggregation involves cell surface glycosaminoglycans.
DeLisser H M, et al.
The Journal of Biological Chemistry, 268(21), 16037-16046 (1993)
Inhibin is a novel paracrine factor for tumor angiogenesis and metastasis
Singh P, et al.
Cancer Research, 78(11), 2978-2989 (2018)
L Piali et al.
The Journal of cell biology, 130(2), 451-460 (1995-07-01)
To protect the body efficiently from infectious organisms, leukocytes circulate as nonadherent cells in the blood and lymph, and migrate as adherent cells into tissues. Circulating leukocytes in the blood have first to adhere to and then to cross the
Bisphenol A alters-hCG and MIF release by human placenta: an in vitro study to understand the role of endometrial cells
Mannelli C, et al.
Mediators of Inflammation, 2014 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service