Skip to Content
Merck
All Photos(1)

Key Documents

BX1715

Supelco

2-Butoxyethanol

Practical, Technical

Synonym(s):

Ethylene glycol butyl ether, Ethylene Glycol Monobutyl Ether, Butoxyethanol, 2-Butoxyethanol, Butyl glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3OCH2CH2OH
CAS Number:
Molecular Weight:
118.17
Beilstein:
1732511
MDL number:
UNSPSC Code:
12352104
EC Index Number:
203-905-0
bp:
169-172.5 °C (lit.)
vapor pressure:
<1 mmHg ( 20 °C)
10 mmHg ( 81 °C)

vapor density

4.1 (vs air)

Quality Level

vapor pressure

<1 mmHg ( 20 °C)
10 mmHg ( 81 °C)

Assay

≥95% (GC)

form

liquid

autoignition temp.

473 °F

expl. lim.

10.6 %

color

APHA: ≤40

refractive index

n20/D 1.419 (lit.)

pH

7 (20 °C in H2O, as aqueous solution)

bp

169-172.5 °C (lit.)

mp

−75 °C (lit.)

transition temp

flash point 67 °C

solubility

900 g/L (completely miscible)

density

0.9 g/cm3 at 20 °C
0.902 g/mL at 25 °C (lit.)

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCOCCO

InChI

1S/C6H14O2/c1-2-3-5-8-6-4-7/h7H,2-6H2,1H3

InChI key

POAOYUHQDCAZBD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (GC): 95% min
Color (APHA): 40 max
Form: Clear liquid
Infrared Spectrum: Conforms to standard

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

152.6 °F - Pensky-Martens closed cup

Flash Point(C)

67 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service