Skip to Content
Merck
All Photos(2)

Key Documents

8.01554

Sigma-Aldrich

Ethylene glycol monobutyl ether

for synthesis

Synonym(s):

Ethylene glycol butyl ether, 2-Butoxyethanol, Butyl glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3OCH2CH2OH
CAS Number:
Molecular Weight:
118.17
Beilstein:
1732511
MDL number:
UNSPSC Code:
12352104
EC Index Number:
203-905-0
NACRES:
NA.22
Assay:
≥99% (GC)
bp:
169-172.5 °C (lit.)
vapor pressure:
<1 mmHg ( 20 °C)
10 mmHg ( 81 °C)
Pricing and availability is not currently available.

vapor density

4.1 (vs air)

Quality Level

vapor pressure

<1 mmHg ( 20 °C)
10 mmHg ( 81 °C)

Assay

≥99% (GC)

form

liquid

autoignition temp.

473 °F

potency

470 mg/kg LD50, oral (Rat)

expl. lim.

10.6 %

refractive index

n20/D 1.419 (lit.)

pH

7 (20 °C in H2O, as aqueous solution)

kinematic viscosity

3.642 cSt(20 °C)

bp

169-172.5 °C (lit.)

mp

−75 °C (lit.)

transition temp

flash point 67 °C

density

0.902 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

CCCCOCCO

InChI

1S/C6H14O2/c1-2-3-5-8-6-4-7/h7H,2-6H2,1H3

InChI key

POAOYUHQDCAZBD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Insight into the aggregation and phase behavior for aqueous solution of ethylene glycol monobutyl ether with self-assembly induced optical and electrical properties: Explores the solubility and amphiphilic properties of ethylene glycol monobutyl ether, affecting its aggregation and phase behavior in solutions (Shang et al., 2023).
  • Surface tension and density of ethylene glycol monobutyl ether–water mixtures from 277.15 to 308.15 K: This study provides detailed measurements of surface tension and density across different temperatures, aiding in the understanding of its physical properties in solutions (Velasco-Medina et al., 2020).
  • Production of ethylene glycol monobutyl ether through etherification of ethylene glycol using a nanostructured heterogeneous catalyst of amyberlyst-15: Describes the synthesis process of ethylene glycol monobutyl ether using advanced catalysis, providing insights into more efficient production techniques (Bayat et al., 2022).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.900 - 0.901
Water (K. F.): ≤ 0.20 %
Peroxide (as H₂O₂): ≤ 0.05 %
Identity (IR): passes test

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

152.6 °F - Pensky-Martens closed cup

Flash Point(C)

67 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service