Skip to Content
Merck
All Photos(3)

Documents

AB765P

Sigma-Aldrich

Anti-Mouse Collagen Type I Antibody

Chemicon®, from rabbit

Synonym(s):

Anti-CAFYD, Anti-EDSC, Anti-OI1, Anti-OI2, Anti-OI3, Anti-OI4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
radioimmunoassay: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... COL1A1(1277)

Related Categories

General description

Collagen Type I extracted and purified from mouse skin. Antibody shows less than 0.1% reactivity with mouse Collagen Types II, IV and with human, rat and chicken Collagens Type I in addition to a 1.0% reactivity with mouse Collagen Type III.
Collagen alpha-1(I) chain (UniProt: P11087; also known as Alpha-1 type I collagen) is encoded by the Col1a1 (also known as Cola1) gene (Gene ID: 12842) in murine species. Collagen is the most abundant protein in the body and make up t 25% of the total protein content. It is predominantly synthesized by fibroblasts. However, epithelial cells may also synthesize small amounts of collagen. The collagen super family includes over 20 different types of collagens with at least 38 distinct polypeptide chains. Collagen type I is a fibrillar type of collagen that is found in skin, tendons, ligaments, and bone and plays a major role in wound healing. Collagen type I is synthesized with a signal peptide (aa 1-22) and a propeptide (aa 23-151) that are subsequently cleaved off to produce the mature form (aa 152-1207). It is expressed in almost all connective tissues and the predominant component of the interstitial membrane. It assembles into fibers that form the structural and mechanical scaffold of bone, skin, tendons, cornea, blood vessel walls and other connective tissues. Heterotrimers of two α1(I) and one α2(I) chains are shown to be the dominant isoform of type I collagen. However, homotrimers of three α1(I) chains have also been reported in fetal tissues, tumors, and in some fibrotic lesions. The homotrimeric isoform is shown to be more resistant to cleavage by collagenases. Type I collagen undergoes several post-translational modifications, some of these are formed during its synthesis to ensure mechanical competence of the fibrils and others formed as a function of aging and disease, such as cleavage and glycation that often results in reduced competence of the fibrils.

Specificity

Antibody shows less than 0.1% reactivity with mouse Collagen Types II, IV and with human, rat and chicken Collagens Type I in addition to a 1.0% reactivity with mouse Collagen Type III.
Recognizes mouse Collagen Type I.

Immunogen

Collagen Type I extracted and purified from mouse skin.

Application

Anti-Mouse Collagen Type 1 is a rabbit polyclonal antibody that detects mouse collagen type 1 and has tested for use in ELISA, Immunohistochemistry, Radioimmunoassay, and Western Blotting.
Research Category
Cell Structure
Research Sub Category
ECM Proteins
Western Blot Analysis:
1:500 dilution of a previous lot detected Collagen Type I on 10 μg of Mouse Liver lysate.

Immunohistochemistry:
1:40 dilution of a previous lot for immunofluorescent staining of frozen mouse skin and liver tissues.

RIA:
A 1:200 dilution of a previous lot was used in RIA.

ELISA:
A previous lot of this antibody was used in ELISA

Target description

MW of Collagen Type I precursor: 138,032 kDa

Linkage

Replaces: AB765

Physical form

Format: Purified
Protein A purified
The purified antibody is supplied in a buffer containing 0.1M Citrate, 0.1M potassium phosphate, at a pH of 7.2-7.4 and 10 µl/ml of antibiotics and antimycotics.

Storage and Stability

Stable for 1 year at -20ºC from date of receipt.

Analysis Note

Control
Mouse skin and mouse liver tissues

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael J Mienaltowski et al.
Stem cell research & therapy, 5(4), 86-86 (2014-07-10)
Multipotent progenitor populations exist within the tendon proper and peritenon of the Achilles tendon. Progenitor populations derived from the tendon proper and peritenon are enriched with distinct cell types that are distinguished by expression of markers of tendon and vascular
Woo-Gyun Choi et al.
Nutrition & metabolism, 14, 48-48 (2017-08-07)
Dietary fructose can rapidly cause fatty liver in animals through de novo lipogenesis (DNL) and contribute to the development and severity of nonalcoholic fatty liver disease (NAFLD). In response to diverse cellular insults including endoplasmic reticulum (ER) and oxidative stress
Glycoprotein VI-dependent and -independent pathways of thrombus formation in vivo.
Dubois, C; Panicot-Dubois, L; Merrill-Skoloff, G; Furie, B; Furie, BC
Blood null
Gerald J Spangrude et al.
Stem cells (Dayton, Ohio), 34(1), 67-82 (2015-10-07)
Splenomegaly is a major manifestation of primary myelofibrosis (PMF) contributing to clinical symptoms and hematologic abnormalities. The spleen from PMF patients contains increased numbers of hematopoietic stem cells (HSC) and megakaryocytes (MK). These MK express high levels of P-selectin (P-sel)
Ming-Hui Fan et al.
The Journal of biological chemistry, 291(15), 8070-8089 (2015-12-15)
Idiopathic pulmonary fibrosis is a disease characterized by progressive, unrelenting lung scarring, with death from respiratory failure within 2-4 years unless lung transplantation is performed. New effective therapies are clearly needed. Fibroblast activation protein (FAP) is a cell surface-associated serine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service