Skip to Content
Merck
All Photos(4)

Key Documents

AB5084P

Sigma-Aldrich

Anti-Dopamine D2 Receptor Antibody

Chemicon®, from rabbit

Synonym(s):

Dopamine D2 receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, monkey, rat, human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... DRD2(1813)
mouse ... Drd2(13489)
rat ... Drd2(24318)
rhesus monkey ... Drd2(694181)

General description

The Dopamine D2 Receptor is one of the five types (D1 to D5) of receptors for dopamine. The activity of this receptor is mediated by G proteins which inhibit adenylyl cyclase. 2 isoforms are reported for the D2 Dopamine Receptor, Isoform 1 and isoform 2 which are expressed in the pituitary and brain; Isoform 1 is expressed seven times more than isoform 2 in the caudate nucleus.

Specificity

Specific for Dopamine Receptor D2 short and long form. Dopamine D2 receptor is known to exist in the short (D2S) and the long (D2L) forms that are encoded by splice variants of a single gene and differ only by the presence of an additional 29 amino acid in the intracellular, cytoplasmic loop 3 (1). D2L receptor is a 446 aa, G-protein coupled and transmembrane receptor protein (2). The immunogen peptide shows no significant homology with other dopamine receptors (D1 and D3-D5).

Immunogen

A 28 amino acid peptide sequence from the human D2 receptor within the cytoplasmic
loop #3.

Application

Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
This Anti-Dopamine D2 Receptor Antibody is validated for use in ELISA, IH, IP, WB for the detection of Dopamine D2 Receptor.
Western blot:
1-10 µg/mL (Chemiluminescence technique) The predicted size of D2 is approximately 50 kDa. However, additional bands at approximately 68 and 100 kDa, and others are typically seen in various tissues (Boundy et al. 1993; Sakata et al. 1992; Farooqui et al. 199; Farooqui & Prasad 1992, BBRC; Brock et al. 1992; Farooqui & Prasad 1992, Life Sci). Approximately 100-400 µg of membrane protein/lane should be loaded to visualize various bands.

Immunohistochemistry:
2-10 μg/mL on paraformaldehyde fixed sections on a previous lot.
Immunohistochemistry:
1-20 µg/mL on paraformaldehyde fixed sections.

Immunoprecipitation:
1-10 μg/mL using 100-400 μg of protein on a previous lot. An antibody made to this epitope has been shown to immunoprecipitate both the long and short forms (2).

ELISA:
0.5-1.0 μg/mL (1 μg/mL immunogen peptide (Catalog number AG221)/well) of a previous lot was used on ELISA.

Optimal working dilutions must be determined by the end user.

Quality

Routinely evaluated by Western Blot on human brain lysates.

Western Blot Analysis: 1:1000 dilution of this lot detected Dopamine Receptor D2 on 10 μg of human brain lysates.

Target description

50 kDa

Physical form

ImmunoAffinity Purified
Purified rabbit polyclonal liquid in PBS with 0.1% BSA.

Storage and Stability

Stable for 1 year at -20ºC from date of receipt.

Analysis Note

Control
Monkey brain tissue, rat brain lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preweaning Mn exposure leads to prolonged astrocyte activation and lasting effects on the dopaminergic system in adult male rats.
Kern CH, Smith DR.
Synapse null
Quantitative, noninvasive, in vivo longitudinal monitoring of gene expression in the brain by co-AAV transduction with a PET reporter gene.
Yoon, SY; Gay-Antaki, C; Ponde, DE; Poptani, H; Vite, CH; Wolfe, JH
Molecular therapy. Methods & clinical development null
Dopamine synthesis and D3 receptor activation in pancreatic ?-cells regulates insulin secretion and intracellular [Ca(2+)] oscillations.
Ustione, A; Piston, DW
Molecular Endocrinology null
Irene Gracia-Rubio et al.
PloS one, 11(12), e0167483-e0167483 (2016-12-10)
Adverse early-life conditions induce persistent disturbances that give rise to negative emotional states. Therefore, early life stress confers increased vulnerability to substance use disorders, mainly during adolescence as the brain is still developing. In this study, we investigated the consequences
Lacey Pflibsen et al.
Journal of neuroscience research, 93(12), 1849-1864 (2015-09-04)
Changes in executive function are at the root of most cognitive problems associated with Parkinson's disease. Because dopaminergic treatment does not necessarily alleviate deficits in executive function, it has been hypothesized that dysfunction of neurotransmitters/systems other than dopamine (DA) may

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service