Skip to Content
Merck
All Photos(1)

Key Documents

AB1792P

Sigma-Aldrich

Anti-Dopamine D2 Receptor Antibody, cytoplasmic domain, long form

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

canine, rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

Specificity

Specific for Dopamine Receptor D2 long form. Dopamine D2 receptor is known to exist in the short (D2S) and the long (D2L) forms that are encoded by splice variants of a single gene and differ only by the presence of an additional 29 amino acid in the intracellular, cytoplasmic loop 3 (Dal Toso et al. 1989; Grandy et al. 1989; Monsma et al. 1990; Giros et al. 1989). D2L receptor is a 446 aa, G-protein coupled and transmembrane receptor protein (Boundy et al. 1993) The 27 aa rat D2RL peptide used for immunization was found only in the D2 long form and there is no significant homology with other dopamine receptors (D1, D2 short, and D3-D5).

SPECIES REACTIVITIES: The immunogen peptide shows sequence homology of 96% with human/bovine (26/27 aa), 62% with Xenopus (17/27 aa) D2L receptors. Antibodies generated to the rat sequence have been shown to recognize the canine D2R (Boundy et al. 1993). Antibody cross-reactivity with various species is not known.

Immunogen

A 27 amino acid peptide sequence from the rat D2 long form within the cytoplasmic loop #3.
Epitope: cytoplasmic domain, long form

Application

Detect Dopamine D2 Receptor using this Anti-Dopamine D2 Receptor Antibody, cytoplasmic domain, long form validated for use in ELISA, IH, IP & WB.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
Western blot: 1-10 μg/mL (Chemiluminescence technique) The predicted size of D2L form is approximately 50 kDa. However, additional bands at approximately 60 and 100 kDa, and others are typically seen in various tissues (Boundy, et al. 1993; Sakata, et al. 1992; Farooqui et al. 1991, 1992; Brock et al. 1992; Farooqui & Prasad 1992). Approximately 100-400 μg of membrane protein/lane should be loaded to visualize various bands.

Immunohistochemistry: 1-20 μg/mL on paraformaldehyde fixed sections.

Immunoprecipitation: 1-10 μg using 100-400 μg of protein (Boundy, et al. 1993).

ELISA: 0.5-1.0 μg/mL (1 μg/mL immunogen peptide (AG211)/well)

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Liquid at 1.0 mg/mL in PBS, pH 7.4 with 0.1% BSA and 0.02% sodium merthiolate.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abu Hasanat Md Zulfiker et al.
Journal of cellular biochemistry, 118(5), 979-993 (2016-06-14)
Recent evidences show that activation of serotonin 2A receptors (5-HT2A R) by agonists is significant in improving therapeutic activity of disease conditions, such as obsessive-compulsive disorder (OCD). Though the exact molecular mechanism is still not well understood, it is thought
Single and binge methamphetamine administrations have different effects on the levels of dopamine D2 autoreceptor and dopamine transporter in rat striatum.
Chauhan, H; Killinger, BA; Miller, CV; Moszczynska, A
International Journal of Molecular Sciences null
M J Drescher et al.
Neuroscience, 171(4), 1054-1074 (2010-10-05)
Adenylyl cyclase (AC) signaling pathways have been identified in a model hair cell preparation from the trout saccule, for which the hair cell is the only intact cell type. The use of degenerate primers targeting cDNA sequence conserved across AC
Jickssa M Gemechu et al.
Scientific reports, 8(1), 1517-1517 (2018-01-26)
Mutations in parkin gene (Park2) are linked to early-onset autosomal recessive Parkinson's disease (PD) and young-onset sporadic PD. Park2 knockout (PKO) rodents; however, do not display neurodegeneration of the nigrostriatal pathway, suggesting age-dependent compensatory changes. Our goal was to examine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service