Skip to Content
Merck
All Photos(1)

Key Documents

8.18652

Sigma-Aldrich

2,4-Dihydroxybenzophenone

for synthesis

Synonym(s):

2,4-Dihydroxybenzophenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2,4-(OH)2-C6H3COC6H5
CAS Number:
Molecular Weight:
214.22
MDL number:
UNSPSC Code:
12352115
EC Index Number:
205-029-4
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥99.0% (acidimetric)

form

powder

potency

>5000 mg/kg LD50, oral (Rat)

pH

5 (20 °C, 10 g/L in H2O, slurry)

mp

143-146 °C

density

1.32 g/cm3 at 20 °C

bulk density

750 kg/m3

storage temp.

2-30°C

SMILES string

Oc1c(ccc(c1)O)C(=O)c2ccccc2

InChI

1S/C13H10O3/c14-10-6-7-11(12(15)8-10)13(16)9-4-2-1-3-5-9/h1-8,14-15H

InChI key

ZXDDPOHVAMWLBH-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 99.0 %
Melting range (lower value): ≥ 143 °C
Melting range (upper value): ≤ 146 °C
Identity (IR): passes test

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

257.0 °F

Flash Point(C)

125 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service