Skip to Content
Merck
All Photos(2)

Key Documents

8.03649

Sigma-Aldrich

N,N′-Diisopropylcarbodiimide

≥99.0% (GC), for peptide synthesis

Synonym(s):

N,N′-Diisopropylcarbodiimide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHN=C=NCH(CH3)2
CAS Number:
Molecular Weight:
126.20
MDL number:
UNSPSC Code:
12352111
EC Index Number:
211-743-7
NACRES:
NA.22

Product Name

N,N′-Diisopropylcarbodiimide, for synthesis

Quality Level

Assay

≥99.0% (GC)

form

liquid

reaction suitability

reaction type: Coupling Reactions

bp

145-148 °C/1013 hPa

transition temp

flash point 33 °C

density

0.81 g/cm3 at 20 °C

application(s)

peptide synthesis

storage temp.

2-30°C

SMILES string

N(=C=NC(C)C)C(C)C

InChI

1S/C7H14N2/c1-6(2)8-5-9-7(3)4/h6-7H,1-4H3

InChI key

BDNKZNFMNDZQMI-UHFFFAOYSA-N

General description

N,N′-Diisopropylcarbodiimide(DIC) is a liquid carbodiimide that contains the functional group RN=C=NR. It is soluble in most organic solvents. DIC is used as a coupling agent in peptide synthesis.

Application

  • Synthesis, structure and reactivity study of magnesium amidinato complexes derived from carbodiimides and N, N′-bis (2, 6-diisopropylphenyl)-1, 4-diaza …: This study involved the preparation of magnesium amidinato complexes using N,N′-Diisopropylcarbodiimide, exploring their structural and reactive properties in various chemical transformations (S Anga, J Bhattacharjee, A Harinath, TK Panda, 2015).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.813 - 0.815
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

91.4 °F

Flash Point(C)

33 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A solvent-reagent selection guide for Steglich-type esterification of carboxylic acids
Jordan A, et al.
Green Chemistry, 23(17), 6405-6413 (2021)
Diisopropylcarbodiimide
Verma A
Synlett, 23(07), 1099-1100 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service