Skip to Content
Merck
All Photos(1)

Key Documents

573099

Sigma-Aldrich

STAT3 Inhibitor V, Stattic

STAT3 Inhibitor V, Stattic, CAS 19983-44-9, is a cell-permeable inhibitor of STAT3 cellular function that targets the STAT3-SH2 domain and prevents its association with upstream Kinases.

Synonym(s):

STAT3 Inhibitor V, Stattic, 6-Nitrobenzo[b]thiophene-1,1-dioxide, Stat three inhibitory compound

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H5NO4S
CAS Number:
Molecular Weight:
211.19
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 10 mg/mL
ethanol: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[S]1(=O)(=O)c2c(ccc(c2)[N+](=O)[O-])C=C1

InChI

1S/C8H5NO4S/c10-9(11)7-2-1-6-3-4-14(12,13)8(6)5-7/h1-5H

InChI key

ZRRGOUHITGRLBA-UHFFFAOYSA-N

General description

A cell-permeable vinyl-sulfone compound that acts as an inhibitor of STAT3 cellular function by targeting the STAT3-SH2 domain and preventing its association with upstream kinases. The effect of Stattic towards STAT family members is temperature-dependent and appears to be highly STAT3-selective at 37°C. Shown to inhibit cellular phosphorylation of STAT3 at Tyr705 with little effect towards STAT1 phosphorylation at Tyr701 (in HepG2 cells) or the phosphorylation of JAK1, JAK2, and c-Src (in MDA-MB-231 and MDA-MB-235S cells).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
STAT3
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C. Note: The use of freshly opened DMSO strongly recommended; the thiol impurities present in DMSO will modify the molecule. Highly recommended to reconstitute just before each use.

Other Notes

Schust, J., et al. 2006. Chem. Biol.13, 1235.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paulraj Kanmani et al.
American journal of physiology. Lung cellular and molecular physiology, 324(5), L596-L608 (2023-03-08)
Inflammasome activation is of central importance for the process of generation of overwhelming inflammatory response and the pathogenesis of sepsis. The intrinsic molecular mechanism for controlling inflammasome activation is still poorly understood. Here we investigated the role of p120-catenin expression
Sivan Izraely et al.
Cells, 12(11) (2023-06-10)
Previous studies from our lab demonstrated that the crosstalk between brain-metastasizing melanoma cells and microglia, the macrophage-like cells of the central nervous system, fuels progression to metastasis. In the present study, an in-depth investigation of melanoma-microglia interactions elucidated a pro-metastatic
Wu Zheng et al.
Journal of neuroinflammation, 19(1), 52-52 (2022-02-20)
Multiple sclerosis (MS) is one of the most common autoimmune disorders characterized by the infiltration of immune cells into the brain and demyelination. The unwanted immunosuppressive side effect of therapeutically successful natalizumab led us to focus on the choroid plexus (CP)
Luigi Ippolito et al.
EMBO reports, 25(8), 3506-3531 (2024-06-22)
Extracellular matrix (ECM) is a major component of the tumor environment, promoting the establishment of a pro-invasive behavior. Such environment is supported by both tumor- and stromal-derived metabolites, particularly lactate. In prostate cancer (PCa), cancer-associated fibroblasts (CAFs) are major contributors
Pauliina Filppu et al.
JCI insight, 6(9) (2021-05-15)
Glioma stem cells (GSCs) drive propagation and therapeutic resistance of glioblastomas, the most aggressive diffuse brain tumors. However, the molecular mechanisms that maintain the stemness and promote therapy resistance remain poorly understood. Here we report CD109/STAT3 axis as crucial for

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service