Skip to Content
Merck
All Photos(1)

Key Documents

422708

Sigma-Aldrich

KN-93

≥95% (HPLC), solid, CAM kinase II inhibitor, Calbiochem®

Synonym(s):

2-[N-(2-hydroxyethyl)]-N-(4-methoxybenzenesulfonyl)]amino-N-(4-chlorocinnamyl)-N-methylbenzylamine)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H29ClN2O4S
CAS Number:
Molecular Weight:
501.04
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

product name

KN-93, A cell-permeable, reversible and competitive inhibitor of rat brain CaM kinase II (Ki = 370 nM).

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C26H29ClN2O4S/c1-28(17-5-6-21-9-11-23(27)12-10-21)20-22-7-3-4-8-26(22)29(18-19-30)34(31,32)25-15-13-24(33-2)14-16-25/h3-16,30H,17-20H2,1-2H3/b6-5+

InChI key

LLLQTDSSHZREGW-AATRIKPKSA-N

General description

A cell-permeable, reversible and competitive inhibitor of rat brain CaM kinase II (Ki = 370 nM). Selectively binds to the CaM binding site of the enzyme and prevents the association of CaM with CaMKII. Has no significant effects on protein kinase A activity. Induces G1 cell cycle arrest and apoptosis in NIH/3T3 cells. A 5 mM (1 mg/399 µl) solution of KN-93 (Cat. No. 422712) in DMSO is also available.
Cell-permeable, reversible, and competitive inhibitor of rat brain Ca+2/calmodulin-dependent protein kinase II (Ki = 370 nM). KN-93 selectively binds to the Calmodulin (CaM) binding site of the enzyme and prevents the association of CaM with CaM Kinase II. Selectively binds to the CaM binding site of the enzyme and prevents the association of CaM with CaMKII. Has no significant effect on protein kinase A activity. Induces G1 cell cycle arrest and apoptosis in NIH 3T3 cells.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
cam kinase 2
Product competes with ATP.
Reversible: yes
Target Ki: 370 nM against rat brain CaM kinase II

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 4 months at -20°C.

Other Notes

Fan, G.H., et al. 1999. Mol. Pharmacol.56, 39.
Tombes, R.M., et al. 1995. Cell Growth Differ.6, 1063.
Mamiya, N., et al. 1993. Biochem. Biophys. Res. Commun. 195, 608.
Nicki, I., et al. 1993. Biochem. Biophys. Res. Commun.191, 255.
Sumi, M., et al. 1991. Biochem. Biophys. Res. Commun.181, 968.
Tokumitsu, H., et al. 1990. J. Biol. Chem.265, 4315.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Siyi Li et al.
Neuroreport, 35(4), 258-268 (2024-02-02)
Diabetic neuropathic pain (DNP) is a frequent complication of diabetes. Calcium/calmodulin-dependent protein kinase II α (CaMKIIα), a multi-functional serine/threonine kinase subunit, is mainly located in the surface layer of the spinal cord dorsal horn (SCDH) and the primary sensory neurons
Xiao-Fen He et al.
Purinergic signalling (2022-01-02)
Diabetic neuropathic pain (DNP) is frequent among patients with diabetes. We previously showed that P2X3 upregulation in dorsal root ganglia (DRG) plays a role in streptozotocin (STZ)-induced DNP but the underlying mechanism is unclear. Here, a rat model of DNP
Tiziana Ravasenga et al.
Cell reports, 38(6), 110347-110347 (2022-02-10)
The induction of synaptic plasticity at an individual dendritic glutamatergic spine can affect neighboring spines. This local modulation generates dendritic plasticity microdomains believed to expand the neuronal computational capacity. Here, we investigate whether local modulation of plasticity can also occur
Bailong Xiao et al.
The Biochemical journal, 396(1), 7-16 (2006-02-18)
We have recently shown that RyR2 (cardiac ryanodine receptor) is phosphorylated by PKA (protein kinase A/cAMP-dependent protein kinase) at two major sites, Ser-2030 and Ser-2808. In the present study, we examined the properties and physiological relevance of phosphorylation of these
Kyoung Mi Sim et al.
Cells, 9(5) (2020-05-03)
ANO1, a Ca2+-activated chloride channel, is highly expressed in glioblastoma cells and its surface expression is involved in their migration and invasion. However, the regulation of ANO1 surface expression in glioblastoma cells is largely unknown. In this study, we found

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service