Skip to Content
Merck
All Photos(1)

Key Documents

420355

Sigma-Aldrich

Isoprenaline hydrochloride

≥99% (TLC), solid, β-adrenoceptor agonist, Calbiochem

Synonym(s):

Isoproterenol, Hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17NO3 · xHCl
CAS Number:
Molecular Weight:
211.26 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

product name

Isoproterenol, Hydrochloride, Phenethylamine derivative.

Quality Level

Assay

≥99% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to off-white
white

solubility

ethanol: soluble
water: soluble

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C11H17NO3.ClH/c1-7(2)12-6-11(15)8-3-4-9(13)10(14)5-8;/h3-5,7,11-15H,6H2,1-2H3;1H

InChI key

IROWCYIEJAOFOW-UHFFFAOYSA-N

General description

Phenethylamine derivative. Selective β-adrenergic agonist that increases cytosolic cAMP. Promotes glycogen breakdown and causes bronchodilation. Increases Cl- diffusion and potential difference in rabbit trachea cells through nitric oxide generation.
Phenethylamine derivative. Selective β-adrenergic agonist that stimulates adenylate cyclase activity. Promotes glycogen breakdown and causes bronchodilation. Increases Cl diffusion and potential difference in rabbit trachea cells through nitric oxide generation. Reported to activate MAP Kinase (ERK) by a Rap1 and B-Raf dependent process.

Biochem/physiol Actions

Cell permeable: no
Primary Target
β-adrenergic agonist
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Irritant (B)

Reconstitution

Unstable in solution; reconstitute just prior to use.

Other Notes

Schmitt, J.M., and Stork, P.J. 2000. J. Biol. Chem.275, 25342.
Carmena, M.J., et al. 1997. Prostate 33, 46.
Takemura, H., et al. 1995. J. Pharmacol. Exp. Ther. 274, 584.
Kishimoto, I., et al. 1994. J. Biol. Chem.269, 28300.
Macaulay, S.L., et al. 1994. Mol. Cell Biochem.141, 27.
Yashiro, K., et al. 1994. J. Biochem.115, 1040.
Ladenheim, R.G., et al. 1993. J. Neurochem.60, 260.
von Zastrow, M., et al. 1993. J. Biol. Chem.268, 763.
Mokhtari, A., et al. 1985. J. Cyclic Nucleotide Protein Phosphorylation Res.10, 213.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Baowen Liu et al.
Frontiers in pharmacology, 11, 1163-1163 (2020-08-28)
Atropine is commonly used to counter the effects of the parasympathetic neurotransmitter acetylcholine on heart rate in clinical practice, such as in the perioperative period; however, individual differences in the response to atropine are huge. The association between SCN10A/voltage-gated sodium
Ivan J Vechetti et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 35(6), e21644-e21644 (2021-05-26)
How regular physical activity is able to improve health remains poorly understood. The release of factors from skeletal muscle following exercise has been proposed as a possible mechanism mediating such systemic benefits. We describe a mechanism wherein skeletal muscle, in

Articles

Gain a deeper understanding of the various types of receptor agonists and antagonists, including full, partial, and inverse agonists and competitive/reversible and non-competitive/irreversible antagonists.

Gain a deeper understanding of the various types of receptor agonists and antagonists, including full, partial, and inverse agonists and competitive/reversible and non-competitive/irreversible antagonists.

Gain a deeper understanding of the various types of receptor agonists and antagonists, including full, partial, and inverse agonists and competitive/reversible and non-competitive/irreversible antagonists.

Gain a deeper understanding of the various types of receptor agonists and antagonists, including full, partial, and inverse agonists and competitive/reversible and non-competitive/irreversible antagonists.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service