Skip to Content
Merck
All Photos(1)

Documents

401414

Sigma-Aldrich

Goat Anti-Rat IgG, H & L Chain Specific Fluorescein Conjugate

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
protect from light

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Immunoaffinity purified goat polyclonal antibody conjugated to fluorescein. Recognizes rat IgG, heavy and light chains.
This Goat Anti-Rat IgG, H & L Chain Specific Fluorescein Conjugate is validated for use in Immunoelectrophoresis, IHC, IF for the detection of Rat IgG, H & L Chain Specific.

Application

Immunoelectrophoresis (see comments)

Immunohistochemistry (1:20-1:40)

Immunofluorescence (1:100, indirect)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Other Notes

Monospecific for rat IgG, heavy and light chains, as determined by immunoelectrophoresis against normal rat serum. The fluorescein:protein ratio is determined on a lot-specific basis; please refer to the certificate of analysis. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer F Pinello et al.
iScience, 27(6), 110146-110146 (2024-06-21)
The ancestral gamete fusion protein, HAP2/GCS1, plays an essential role in fertilization in a broad range of taxa. To identify factors that may regulate HAP2/GCS1 activity, we screened mutants of the ciliate Tetrahymena thermophila for behaviors that mimic Δhap2/gcs1 knockout
Anders D Skjolding et al.
Cerebrospinal fluid research, 7, 20-20 (2010-11-09)
The water channel protein aquaporin-4 (AQP4) is reported to be of possible major importance for accessory cerebrospinal fluid (CSF) circulation pathways. We hypothesized that changes in AQP4 expression in specific brain regions correspond to the severity and duration of hydrocephalus.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service