Skip to Content
Merck
All Photos(1)

Key Documents

345717-M

Millipore

Ganglioside Mixture, Ammonium Salt, Bovine Brain

Contains a mixture of GM1, GD1a, GD1b, GT1b, and other gangliosides.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211

Quality Level

Assay

≥98% (TLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

chloroform: methanol: water (2:1:0.1): soluble
ethanol: soluble

storage temp.

−20°C

General description

Contains a mixture of GM1, GD1a, GD1b, GT1b, and other gangliosides.
Extracted from bovine gray matter. Contains a mixture of GM1, GD1a, GD1b, GT1b, and other gangliosides. Useful as a substrate for sialidases.



Note: This product is not to be used for animal treatment, in vivo research or in any other contact procedure with livestock.

Biochem/physiol Actions

Primary Target
Contains a mixture of GM1, GD1a, GD1b, GT1b, and other gangliosides
Product does not compete with ATP.

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service