Skip to Content
Merck
All Photos(1)

Key Documents

238732

Sigma-Aldrich

Ammonium trifluoroacetate

98%

Synonym(s):

Trifluoroacetic acid ammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CO2NH4
CAS Number:
Molecular Weight:
131.05
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

crystals

mp

123-125 °C (lit.)

SMILES string

N.OC(=O)C(F)(F)F

InChI

1S/C2HF3O2.H3N/c3-2(4,5)1(6)7;/h(H,6,7);1H3

InChI key

YCNIBOIOWCTRCL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium Trifluoroacetate is used as a catalyst in organic synthesis and as an additive in the mobile phase for chiral racemate separation.

Application

Used in the synthesis of hexafluoro-2-aminopentan-4-one ligand and in tuning and calibrating new liquid chromatography/mass spectrometry systems.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of Fluorine Chemistry, 27, 371-371 (1985)
Nimisha Thakur et al.
Chirality, 31(9), 688-699 (2019-07-19)
The enantiomeric excess of chiral starting materials is one of the important factors determining the enantiopurity of products in asymmetric synthesis. Fifty-one commercially available chiral reagents used as building blocks, catalysts, and auxiliaries in various enantioselective syntheses were assayed for
Garrett Hellinghausen et al.
Chirality, 30(9), 1067-1078 (2018-07-04)
A modified macrocyclic glycopeptide-based chiral stationary phase (CSP), prepared via Edman degradation of vancomycin, was evaluated as a chiral selector for the first time. Its applicability was compared with other macrocyclic glycopeptide-based CSPs: TeicoShell and VancoShell. In addition, another modified
Ammonium Trifluoroacetate-Mediated Synthesis of 3, 4-dihydropyrimidin-2 (1H)-ones
Chandran R et al.
International Scholarly Research Network, 2011 (2011)
Garrett Hellinghausen et al.
Journal of pharmaceutical and biomedical analysis, 155, 70-81 (2018-04-07)
Core-shell particles (superficially porous particles, SPPs) have been proven to provide high-throughput and effective separations of a variety of chiral molecules. However, due to their limited commercialization, many separations have not been reported with these stationary phases. In this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service