Skip to Content
Merck
All Photos(1)

Key Documents

317496

Sigma-Aldrich

Dual Specificity Protein Phosphatase 1/6 Inhibitor, BCI

The Dual Specificity Protein Phosphatase 1/6 Inhibitor, BCI controls the biological activity of Dual Specificity Protein Phosphatase 1/6. Primarily used for Phosphorylation & Dephosphorylation apps.

Synonym(s):

Dual Specificity Protein Phosphatase 1/6 Inhibitor, BCI, NSC 150117, (E)-2-Benzylidene-3-(cyclohexylamino)-2,3-dihydro-1H-inden-1-one, MAP Kinase Phosphatase 1/3 Inhibitor, BCI, Dusp1/MKP1 Inhibitor, Dusp6/MKP3 Inhibitor, PTP Inhibitor XXVI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H23NO
CAS Number:
Molecular Weight:
317.42
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

N(C4CCCCC4)C1c2c(cccc2)C(=O)\C\1=C/c3ccccc3

InChI

1S/C22H23NO/c24-22-19-14-8-7-13-18(19)21(23-17-11-5-2-6-12-17)20(22)15-16-9-3-1-4-10-16/h1,3-4,7-10,13-15,17,21,23H,2,5-6,11-12H2/b20-15-

General description

A cell-permeable cyclohexylamino-indenone compound that acts as an allosteric inhibitor against substrate binding-induced MAPK phosphatase activity of Dusp1/CL100/HVH1/MKP1/PTPN10 and Dusp6/MKP3/PYST1 (57% and 30% reduction, respectively, of 0.21 and 2.1 µg Erk2-induced OMFP dephosphorylation by 0.21 µg Dusp6) by locking Dusp1 and Dusp6 in their low-activity conformation, while exhibiting little activity toward Cdc25B, DUSP3/VHR, DUSP5, or PTP1B. Shown to effectively enhance cellular Erk phosphorylation in HeLa cells overexpressing human Dusp1 and Dusp6 (EC50 = 11.5 and 12.3 µM, respectively). FGF signaling activation by BCI treatment (5 to 10 µM) during zebrafish embryo development is reported to expand myocardial progenitors population at the expense of other progenitor cell types, resulting in enlarged heart size.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstiution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Molina, G., et al. 2009. Nat. Chem. Biol.5, 680.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sabrina Blumer et al.
Cells, 10(11) (2021-11-28)
Idiopathic pulmonary fibrosis (IPF) is a progressive terminal lung disease, and therapies aim to block fibrosis. Fibroblast proliferation is controlled by C/EBP-β, microRNA cluster 17-92 (miR17-92), and Erk1/2 mitogen-activated protein kinase. This study assessed the role of miR17-92 in IPF-fibroblast
Briana C Prager et al.
Cancer discovery, 10(11), 1722-1741 (2020-07-25)
Meningiomas are the most common primary intracranial tumor with current classification offering limited therapeutic guidance. Here, we interrogated meningioma enhancer landscapes from 33 tumors to stratify patients based upon prognosis and identify novel meningioma-specific dependencies. Enhancers robustly stratified meningiomas into

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service