Skip to Content
Merck
All Photos(3)

Key Documents

S7067

Sigma-Aldrich

SB 202190

≥98% (HPLC), powder, p38 MAP kinase inhibitor

Synonym(s):

4-(4-Fluorophenyl)-2-(4-hydroxyphenyl)-5-(4-pyridyl)-1H-imidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H14FN3O
CAS Number:
Molecular Weight:
331.34
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

SB 202190, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

color

faintly yellow, to beige

solubility

DMSO: 10 mg/mL, clear

originator

GlaxoSmithKline

storage temp.

2-8°C

SMILES string

Oc1ccc(cc1)-c2nc(-c3ccc(F)cc3)c([nH]2)-c4ccncc4

InChI

1S/C20H14FN3O/c21-16-5-1-13(2-6-16)18-19(14-9-11-22-12-10-14)24-20(23-18)15-3-7-17(25)8-4-15/h1-12,25H,(H,23,24)

InChI key

QHKYPYXTTXKZST-UHFFFAOYSA-N

Gene Information

human ... MAPK14(1432)

General description

SB202190 aids in enhancing the self-renewal ability of neural stem cells fromNPC1-deficient mice.

Application

SB 202190 has been used:
  • to investigate the role of p38mitogen-activated protein kinase (MAPK) signaling in establishment of reovirus infection
  • as a p38α inhibitor in in vivo studies to study its impact on the development or progression of adenocarcinoma in the colon mucosa tissues of APC Min/+ mice.
  • as a component of basic BC medium to culture basal progenitor cells.
  • as a component of normal endometrium and endometrial tumor media for organoid culture.
SB 202190 was used to inhibit p38 activation in MCF7 cells5, mouse macrophages6 and HepG2 cells.7

Biochem/physiol Actions

SB 202190 is a highly selective, potent and cell permeable inhibitor of p38 MAP kinase. SB 202190 binds within the ATP pocket of the active kinase (Kd = 38 nM, as measured in recombinant human p38), and selectively inhibits the p38α and β isoforms.

Features and Benefits

This compound is featured on the MAPKs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Legal Information

Sold for research purposes under agreement from Glaxo­Smith­Kline

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

NPC1 gene deficiency leads to lack of neural stem cell self-renewal and abnormal differentiation through activation of p38 mitogen-activated protein kinase signaling
Yang SR, et al.
Journal of Separation Science, 24(2), 292-298 (2006)
Uncoupling p38? nuclear and cytoplasmic functions and identification of two p38? phosphorylation sites on ?-catenin: implications for the Wnt signaling pathway in CRC models
Lepore Signorile M, et al.
Journal of Separation Science, 13(1), 223-223 (2023)
Protocol for generating airway organoids from 2D air liquid interface-differentiated nasal epithelia for use in a functional CFTR assay
Rodenburg LW, et al.
STAR protocols, 4(3), 102337-102337 (2023)
Edward Thorp et al.
The Journal of biological chemistry, 286(38), 33335-33344 (2011-08-11)
Mer tyrosine kinase (MerTK) is an integral membrane protein that is preferentially expressed by phagocytic cells, where it promotes efferocytosis and inhibits inflammatory signaling. Proteolytic cleavage of MerTK at an unidentified site leads to shedding of its soluble ectodomain (soluble
Shauna M Crowley et al.
PLoS pathogens, 16(4), e1008498-e1008498 (2020-04-14)
We investigated the role of the inflammasome effector caspases-1 and -11 during Salmonella enterica serovar Typhimurium infection of murine intestinal epithelial cells (IECs). Salmonella burdens were significantly greater in the intestines of caspase-1/11 deficient (Casp1/11-/-), Casp1-/- and Casp11-/- mice, as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service