Skip to Content
Merck
All Photos(1)

Documents

256455-M

Millipore

Big CHAP, Deoxy

A non-ionic detergent analog of CHAPS and CHAPSO. Has reduced electrostatic interactions that do not interfere in anion exchange chromatography on DEAE-cellulose. Aggregation number: 8-16.

Synonym(s):

Big CHAP, Deoxy, N,N- bis-(3-D-Gluconamidopropyl)deoxycholamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H75N3O15
CAS Number:
Molecular Weight:
862.06
UNSPSC Code:
12161900
NACRES:
NA.25

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

aggregation number

8 - 16

color

white

CMC

1.1 - 1.4 mM

solubility

water: 5%

storage temp.

10-30°C

General description

A non-ionic detergent analog of CHAPS and CHAPSO. Has reduced electrostatic interactions that do not interfere in anion exchange chromatography on DEAE-cellulose. Aggregation number: 8-16.
A non-ionic detergent analog of CHAPS and CHAPSO. Has reduced electrostatic interactions that do not interfere in anion exchange chromatography on DEAE-cellulose. Has a critical micelle concentration (CMC) of 1.1-1.4 mM and an aggregation number of 8-16.

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 6 months at 4°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service