Skip to Content
Merck
All Photos(3)

Key Documents

189898

Sigma-Aldrich

Methylmagnesium bromide solution

3.0 M in diethyl ether

Synonym(s):

Bromomethylmagnesium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3MgBr
CAS Number:
Molecular Weight:
119.24
Beilstein:
3535220
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

Quality Level

reaction suitability

reaction type: Grignard Reaction

concentration

3.0 M in diethyl ether

density

1.035 g/mL at 25 °C

SMILES string

C[Mg]Br

InChI

1S/CH3.BrH.Mg/h1H3;1H;/q;;+1/p-1

InChI key

AVFUHBJCUUTGCD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Methylmagnesium bromide solution, also known as bromomethylmagnesium, is both a Lewis acid and nucleophile. It is commonly used as a grignard reagent in organic synthesis reactions.

Application

Methylmagnesium bromide solution (3M in diethyl ether) is used to synthesize:
  • 3S-(Boc-amino)-2,4-dimethyl-2-pentanol and methyl tert-butyl (S)-sulfoxide.
  • Methyl potassium polyimide (K-PIM-1) (MeMgBr).
  • 3(4R)-{3(2R,3S)-[4′-(1′-triphenylmethyl)imidazole]-2-bromo-1-oxobutyl)-4-phenyl2-oxazolidinone
  • Benzyl 2-(2-oxopropyl)piperidine-1-carboxylate
  • (R*, R*) 1-[1-(1-phenylethyl)pyrrolidin-2-yl]propan-2-one)

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 1 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric synthesis of (2R, 3S) and (2S, 3R) precursors of ?-methyl-histidine,-phenylalanine and-tyrosine.
Li G
Tetrahedron Asymmetry, 4(11), 2315-2318 (1993)
Mechanistic insights on the stereoselective nucleophilic 1, 2-addition to sulfinyl imines
Martin H,et al.
The Journal of Organic Chemistry, 79, 2514-2521 (2014)
Diastereoselective preparation of cyclic ?-aminoketals. Application to the total synthesis of piperidine alkaloids
Al-Sarabi AE
ARKIVOC (Gainesville, FL, United States), 1, 119-133 (2007)
DMAP-Catalysed Sulfinylation of Diacetone-D-Glucose: Improved Method for the Synthesis of Enantiopure tert-Butyl Sulfoxides and tert-Butanesulfinamides
Ahmed C,et al.
European Journal of Organic Chemistry, 2014, 6935-6944 (2014)
Organometallic Complexes of Bulky, Optically Active, C 3-Symmetric Tris (4 S-isopropyl-5, 5-dimethyl-2-oxazolinyl) phenylborate (ToP*)
Songchen X,et al.
Organometallics, 34, 3508-3519 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service