Skip to Content
Merck
All Photos(1)

Key Documents

1.93237

Supelco

Sodium hydrogen carbonate

for analysis EMPARTA® ACS

Synonym(s):

Sodium bicarbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaHCO3
CAS Number:
Molecular Weight:
84.01
Beilstein:
4153970
MDL number:
UNSPSC Code:
12352302
grade:
ACS reagent
Pricing and availability is not currently available.

grade

ACS reagent

pKa (25 °C)

(1) 6.37, (2) 10.25 (carbonic acid)

density

2.16 g/mL at 25 °C (lit.)

storage temp.

room temp

SMILES string

[Na+].OC([O-])=O

InChI

1S/CH2O3.Na/c2-1(3)4;/h(H2,2,3,4);/q;+1/p-1

InChI key

UIIMBOGNXHQVGW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Packaging

1.93237.0521: 500 g, Minimum order quantity: 20
1.93237.9051: 50 kg, Minimum order quantity: 1

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Inorganics and solvents for basic and standard applications manufactured locally.

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service