Skip to Content
Merck
All Photos(1)

Key Documents

1.93003

Supelco

ortho-Phosphoric acid 88%

for analysis EMPARTA® ACS

Synonym(s):

Phosphoric acid, Orthophosphoric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3PO4
CAS Number:
Molecular Weight:
98.00
Beilstein:
1921286
MDL number:
UNSPSC Code:
12352106
Pricing and availability is not currently available.

grade

ACS reagent

vapor density

3.4 (vs air)

vapor pressure

2.2 mmHg ( 20 °C)
5 mmHg ( 25 °C)

product line

EMPARTA®

bp

158 °C (lit.)

mp

~40 °C (lit.)

density

1.685 g/mL at 25 °C (lit.)

storage temp.

room temp

SMILES string

OP(O)(O)=O

InChI

1S/H3O4P/c1-5(2,3)4/h(H3,1,2,3,4)

InChI key

NBIIXXVUZAFLBC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

1.93003.0521: 500ML, Minimum order quantity: 20
1.93003.2521: 2.5 L, Minimum order quantity: 4
1.93003.5021: 5 L, Minimum order quantity: 4
1.93003.9026: 25 L, Minimum order quantity: 1

Other Notes

Concentrated phosphoric acid is approx. 85% H3PO4 (aqueous).

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Inorganics and solvents for basic and standard applications manufactured locally.

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service