Skip to Content
Merck
All Photos(1)

Key Documents

1.10784

Millipore

Acrylamide

for electrophoresis

Synonym(s):

Acrylamide, Prop-2-enamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5NO
CAS Number:
Molecular Weight:
71.08
MDL number:
UNSPSC Code:
41105319
EC Index Number:
201-173-7
NACRES:
NB.21
Pricing and availability is not currently available.

vapor pressure

0.009 hPa ( 25 °C)

Quality Level

form

solid

autoignition temp.

424 °C

potency

177 mg/kg LD50, oral (Rat)
1141 mg/kg LD50, skin (Rabbit)

pH

5.0-7.0 (20 °C, 50 g/L in H2O)

bp

125 °C/33.3 hPa

mp

84 °C

transition temp

flash point 138 °C

solubility

2040 g/L

density

1.127 g/cm3 at 25 °C

bulk density

500 kg/m3

storage temp.

2-30°C

SMILES string

NC(=O)C=C

InChI

1S/C3H5NO/c1-2-3(4)5/h2H,1H2,(H2,4,5)

InChI key

HRPVXLWXLXDGHG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Direct 3D printing of triple-responsive nanocomposite hydrogel microneedles for controllable drug delivery.: This research develops novel 3D-printed hydrogel microneedles using acrylamide as a component that responds to environmental changes (Li H et al., 2024).

Analysis Note

Appearance (colour): almost white
Appearance (description): crystalline powder
Assay (GC): ≥ 99.9 %
Identity (IR-spectrum): passes test
Free acid (as acrylic acid): ≤ 0.001 %
Absorbance A (290 nm; 1 %; 1 cm; water): ≤ 0.15
Conductivity (50 % w/v; water; 20° C): ≤ 5.0 µS/cm
Solubility (50 % w/v; water): clear, colourless
Turbidity (50 % w/v; water): ≤ 2.0 NTU
Turbidity (50 % w/v; methanol; 37° C): ≤ 3.0 NTU
pH-value (5 %; water): 5.0 - 7.0
Water-insoluble matter: ≤ 0.005 %

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Oral

Target Organs

Peripheral nervous system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

280.4 °F - closed cup

Flash Point(C)

138 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service