Skip to Content
Merck
All Photos(1)

Key Documents

1.08342

Millipore

D-(+)-Glucose monohydrate

suitable for microbiology, pH 6-7 (20 °C, 100 g/L in H2O)

Synonym(s):

Dextrose, , Dextrose monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6 · H2O
CAS Number:
Molecular Weight:
198.17
Beilstein:
5250278
MDL number:
UNSPSC Code:
41106212
EC Index Number:
200-075-1
NACRES:
NA.74
Pricing and availability is not currently available.

form

solid

Quality Level

autoignition temp.

500 °C

potency

25800 mg/kg LD50, oral (Rat)

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 1 kg
pkg of 2.5 kg
pkg of 25 kg

pH

6-7 (20 °C, 100 g/L in H2O)

mp

83 °C

bulk density

630 kg/m3

application(s)

microbiology

storage temp.

15-25°C

SMILES string

O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C=O

InChI

1S/C6H12O6.H2O/c7-1-3(9)5(11)6(12)4(10)2-8;/h1,3-6,8-12H,2H2;1H2/t3-,4+,5+,6+;/m0./s1

InChI key

SPFMQWBKVUQXJV-BTVCFUMJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

D-(+)-Glucose monohydrate, commonly known as glucose or dextrose, has several microbiological applications. It is used as a carbon source in microbial culture media for the growth and propagation of a wide range of microorganisms. Glucose is a simple sugar and a primary source of energy for bacterial cell metabolism. Glucose monohydrate can also be used as a supplement in media to enhance the growth and productivity of microorganisms used in fermentation processes, for example, in the production of antibiotics, organic acids, and enzymes. It plays an important role in diauxic growth, which refers to the phenomenon in which bacterial cells preferentially utilize glucose as a carbon source before switching to other energy sources. In addition, glucose is a common component in numerous selective and differential media formulations used in the identification and characterization of microorganisms.

Application

D-(+)-Glucose monohydrate is commonly used as an ingredient in many microbial culture media for the growth and identification of bacteria, yeasts and molds from various pharmaceutical, environmental, food and beverage samples.

Analysis Note

Identity (IR-spectrum): passes test
Appearance of solution (10 %; water): passes test
Spec. rotation (α 20/D; 10 %; water; calc. on anhydrous substance): +52.5 - +53.2 °
Heavy metals (as Pb): ≤ 0.0005 %
Maltose (HPLC): ≤ 0.2 %
Water: 8 - 10 %
Suitability for microbiology: passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sina V Barysch et al.
Nature protocols, 5(6), 1127-1137 (2010-06-12)
Early endosomes receive material from the plasma membrane by fusion with endocytotic vesicles. This material is sorted within endosomes and directed to subdomains at which carrier vesicles bud. These vesicles are then transported toward different cellular destinations. In this article

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service