Skip to Content
Merck
All Photos(1)

Key Documents

1.08337

Millipore

D-(+)-Glucose

anhydrous for biochemistry Reag. Ph Eur

Synonym(s):

Dextrose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1724615
MDL number:
UNSPSC Code:
12352201
EC Index Number:
200-075-1
NACRES:
NA.79
Pricing and availability is not currently available.

Agency

reag. Ph. Eur.

Quality Level

form

solid

autoignition temp.

500 °C

potency

25800 mg/kg LD50, oral (Rat)

mp

150-152 °C (lit.)

solubility

470 g/L

bulk density

630 kg/m3

storage temp.

15-25°C

SMILES string

OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1

InChI key

WQZGKKKJIJFFOK-DVKNGEFBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

anhydrous for biochemistry Reag. Ph Eur

Application

D(+)-Glucose anhydrous for biochemistry Reag. Ph Eur. CAS 50-99-7, molar mass 180.16 g/mol.

Analysis Note

Assay (calc. on anhydrous substance)97.5 - 102.0 %Identity (specific rotation)passes testIdentity (HPLC)passes testIdentity (water)passes testAppearance of solutionpasses testConductivity≤ 20 µS/cmRelated substances (sum of impurities A and B)≤ 0.4 %Related substances (impurity C)≤ 0.2 %Related substances (impurity D)≤ 0.15 %Related substances (unspecified impurities)≤ 0.10 %Related substances (Sum of all impurities)≤ 0.5 %Dextrinpasses testSoluble starch, sulfite≤ 15 ppmResidual solvents (ICH Q3C)excluded by the manufacturing processWater (according to Karl Fischer)≤ 1.0 %Corresponds to Reag. Ph.Eur.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service