0796
Buffered Charcoal Yeast Extract (BCYE) Agar

ready-to-use, plate of 20 × 23 ± 1 mL, selective for bacteria: Legionella spp
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106217
Recommended Products
Agency
ISO 11731
form
plate (Semi-solid Media Filled)
feature
ready-to-use
packaging
plate of 20 × 23 ± 1 mL
technique(s)
microbiological culture: suitable
plate size
90 mm
pH
6.9 ± 0.2 (25 °C)
application(s)
food and beverages
water monitoring
storage temp.
2-8°C
suitability
selective for bacteria: Legionella spp
General description
BCYE Agar, also known as Buffered Charcoal Yeast Extract Agar, is a Ready-to-Use media designed as per ISO Standards 11731 to detect Legionella spp.
The media contains charcoal, which serves as a detoxifying agent. Yeast extract provides a rich source of vitamins, nitrogen, and carbon. ACES Buffer helps maintain an optimal pH for growth, while L-cystine hydrochloride, ferric pyrophosphate, and α-ketoglutarate stimulate the growth of Legionella species.
Legionella a gram-negative, rod-shaped bacteria, commonly found in various freshwater biotopes, often associated with protozoans such as amoebae, where they can grow intracellularly. Legionella causes pneumonia (Legionnaires disease) and febrile disease (Pontiac fever)
The media contains charcoal, which serves as a detoxifying agent. Yeast extract provides a rich source of vitamins, nitrogen, and carbon. ACES Buffer helps maintain an optimal pH for growth, while L-cystine hydrochloride, ferric pyrophosphate, and α-ketoglutarate stimulate the growth of Legionella species.
Legionella a gram-negative, rod-shaped bacteria, commonly found in various freshwater biotopes, often associated with protozoans such as amoebae, where they can grow intracellularly. Legionella causes pneumonia (Legionnaires disease) and febrile disease (Pontiac fever)
Application
Buffered Charcoal Yeast Extract Agar is used to enumerate, detect and isolate Legionella spp in water samples from food and beverage industry.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service